Skip to Content
Merck
All Photos(1)

Documents

01-6570

Sigma-Aldrich

Arsenic standard solution

1 mg/mL As, 1000 ppm As, suitable for atomic absorption spectrometry

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

form

liquid

availability

available only in Japan

concentration

1 mg/mL As
1000 ppm As

suitability

suitable for atomic absorption spectrometry

storage temp.

15-25°C

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service