Skip to Content
Merck
All Photos(1)

Key Documents

RIEH2F64B0BF

Rieke Metals, Inc.

5-Methoxy-2-pyridylzinc bromide 0.5 M in Tetrahydrofuran

0.5 M in Tetrahydrofuran

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
253.41
MDL number:

Iupac Name

(5-methoxypyridin-2-yl)zinc(II) bromide

Inchi Code

1S/C6H6NO.BrH.Zn/c1-8-6-3-2-4-7-5-6;;/h2-3,5H,1H3;1H;/q;;+1/p-1

InChI key

DEQMCEKJXSETDA-UHFFFAOYSA-M

Country of Origin

US

Physical Form

Liquid

Product Link

https://riekemetals.com/products-services/catnum/2406

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library