Skip to Content
Merck
All Photos(1)

Key Documents

ST1543

Sigma-Aldrich

Anti-GHR Mouse mAb (3A12)

liquid, clone 3A12, Calbiochem®

Synonym(s):

Anti-Growth Hormone Receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 371.00

CHF 371.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 371.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 371.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3A12, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GHR(2690)

General description

Anti-GHR, mouse monoclonal, clone 3A12, recognizes the GHR protein in HeLa cells. It is validated for use in ELISA, Western blotting, and immunocytochemistry.
Purified mouse monoclonal antibody. Recognizes the GHR protein.
Recognizes the GHR protein in HeLa cells.

Immunogen

A recombinant polypeptide corresponding to amino acids 19-119 of human GHR, expressed as a GST fusion protein

Application



ELISA (see coments)
Immunoblotting (1-5 g/ml)
Immunocytochemistry (1:50)

Warning

Toxicity: Regulatory Review (Z)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service