Skip to Content
Merck
All Photos(1)

Documents

SCM05

Sigma-Aldrich

AOF ITS Supplement (100X), 100ml | SCM055

AOF ITS supplement is an animal-origin-free product that contains human recombinant insulin, transferrin along with sodium selenite and is used for the serum-free culture of numerous cell types.

Synonym(s):

ITS, Insulin Transferrin Selenite

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352205
eCl@ss:
32160801
NACRES:
NA.75

Quality Level

form

liquid

concentration

100 ×

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

General description

EMD Millipore’s AOF ITS (100X) supplement is an animal-origin-free product that contains human recombinant insulin, human recombinant transferrin along with sodium selenite.

Undefined animal derived media supplements, such as fetal bovine serum, have recently been an area of concern for many cell culturists due to their increased costs, lot to lot variability and increased regulation worldwide. The elimination of fetal bovine serum from cell culture media provides a more defined enviroment to study basic biological questions. Serum-free and reduced serum formulations often require purified forms of the factors normally supplied by serum that are suitable for in vitro growth and differentiation studies such as insulin, transferrin and selenite.

ITS supplements have been proven to replace serum and grow the following cells in serum free or reduced serum conditions: CHO, HEK293, fibroblasts, hybridoma, VERO cells, Hela, HT29, MDCK, Mesenchymal, IPS Cells, Cardiac, kidney, keratinocyte, embryonic, neural stem cell, hepatocytes.

Application

EMD Millipore’s AOF ITS (100X) supplement is an completely animal-origin-free product
Research Category
Stem Cell Research

Components

All components are derived using AOF (Animal-Origin-Free) reagents and protocols.

Recombinant Human Insulin is derived from Pichia pastoris (yeast).
Recombinant Human Transferrin is derived from Oryza sativa (rice).
Sodium Selenite is a synthetic chemical.

Quality

Tested for the ability to support the proliferation of VERO cells in serum free conditions over a 3 day time period.
pH: 7.6-8.0.
Product is sterile.
Endotoxin levels are ≤ 1 EU/mg.

Physical form

Product is made in an Earle′s Balanced Salt Solution and filtered through a 0.22 micron membrane before vialing. Solution is a light pink clear solution that is free of particulates.

Storage and Stability

Store at 2-8°C. Further dilutions should be made into appropriate buffer and/or medium to which protein has been added, such as DMEM/1% Human Serum Albumin or equivalent.

Analysis Note

Control
VERO cells

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service