Skip to Content
Merck
All Photos(1)

Documents

S8000-M

Sigma-Aldrich

CpGenome Universal Methylated Mouse DNA Standard Set

The CpGenome Universal Methylated Mouse DNA Standard contains universally methylated mouse genomic DNA isolated from male Balb/c mice & enzymatically methylated at all CpGs by M.SssI methyltransferase.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.31

Quality Level

species reactivity

mouse

manufacturer/tradename

CpGenome
Upstate®

General description

The CpGenome Universal Methylated Mouse DNA Standard contains universally methylated mouse genomic DNA isolated from male Balb/c mice and enzymatically methylated at all CpGs by M.SssI methyltransferase. It can be used as a positive control for gene methylation studies, and is fully compatible with the CpGenome Turbo Bisulfite kit (Cat. No. S7847) and CpG WIZ methylation-specific amplification systems.

Materials Provided:

One vial containing 5 µg (20 µl) of CpGenome Universal Methylated Mouse DNA Standard at a concentration of 250 ng/µL.

Validation:

Methylation-specific PCR (MSP) was performed on the CpGenome Universal Methylated Mouse DNA after bisulfite modification with the CpGenome Turbo Bisulfite kit (Cat. No. S7847). Three sets of primers from the CpG WIZ Oct-4 kits (Catalog No. S7840) were used in this assay: the U primer set, which anneals to unmethylated bisulfite-modified DNA; M primers, which anneal to methylated bisulfite-modified sequences; and W primers, which anneal to unmethylated or methylated DNA that has not undergone bisulfite modification. Only the M primers generated product for this Standard.

CpGenome and CpG WIZ are trademarks of Serologicals Corporation. CpG WIZ Methylation Products apply to technologies exclusively licensed from The Johns Hopkins University School of Medicine. Methylation-specific PCR (MSP) technology is covered under U.S. Patent # 5,786,146.

Application

Research Category
Epigenetics & Nuclear Function

Physical form

Liquid in buffer containing 10 mM Tris-HCl, 1 mM EDTA, pH 8.0

Storage and Stability

Recommended Storage: Stable at -20°C for up to 6 months from date of receipt.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service