Skip to Content
Merck
All Photos(1)

Key Documents

S384SEQ

Millipore

MultiScreen SEQ384 Filter Plate

Dye terminator removal

Synonym(s):

Montage SEQ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.15

material

polystyrene

Quality Level

sterility

non-sterile

manufacturer/tradename

MultiScreen®

parameter

100 μL sample volume (per well)

plate size

384 wells

pore size

pore sizeproprietary

application(s)

sample preparation

General description

Filter Code: SEQ

Application

Research Category
Assays

General Filtration

Nucleic acid

Features and Benefits

Proprietary membrane

Components

Does not include reagents

Physical form

Color Code: Clear

Other Notes

Automation Compatible: No

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service