R03060
Haemalum Stain
in accordance with BSC, contains no acetic acid
Synonym(s):
Counterstain
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116121
Agency
in accordance with BSC
Quality Level
does not contain
acetic acid
shipped in
ambient
storage temp.
10-30°C
General description
For counterstaining in cytochemistry. Mayer`s (without Acetic Acid)
Mayer′s Haemalum Stain
R03060-74 500ml Poly Bottle
R03060-74 500ml Poly Bottle
Application
For counterstaining in cytochemistry. Mayer`s (without Acetic Acid)
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
For counterstaining in cytochemistry.
For In-Vitro Diagnostic Use.
For Professional Use.
For counterstaining in cytochemistry.
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service