Skip to Content
Merck
All Photos(1)

Documents

PDMA04700

Millipore

PetriSlide dish

47mm dish preloaded with adsorbent pads for microbial application using membrane filtration.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122101
eCl@ss:
32030209
NACRES:
NB.14

material

cellulose pad(s)
clear device
polystyrene

Quality Level

sterility

sterile

packaging

plate of 100

manufacturer/tradename

Petrislides

technique(s)

sterile filtration: suitable

diam.

47 mm

input

sample type food(s)
sample(s) (environmental)
sample type beverage(s)
sample(s) (environmental)

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

shipped in

ambient

Related Categories

General description

PetriSlide for Contamination Analysis includes a transparent cover allowing microscopic examination without removal. Rectangular base has rounded corners for mounting on microscope stage. PetriSlide with absorbent pad is a single use disposable plastic unit to be used in culturing microorganisms that has been captured on a 47mm filter by membrane filtration method.

Application

PetriSlide with absorbent pad is a single use disposable plastic unit to be used in microbiological analysis of Beer, Bottled Water, Cider, Sport Drink, Wine etc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service