Skip to Content
Merck
All Photos(1)

Key Documents

OP143

Sigma-Aldrich

Anti-MDM2 (Ab-3) Mouse mAb (4B11)

liquid, clone 4B11, Calbiochem®

Synonym(s):

Anti-Ubiquitin Protein Ligase, Anti-p53 Binding Protein, Anti-Murine Double Minute Chromosome-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 412.00

CHF 412.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 412.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 412.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4B11, monoclonal

form

liquid

does not contain

preservative

species reactivity

mouse, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MDM2(4193)
mouse ... Mdm2(17246)

General description

Purified mouse monoclonal antibody. Recognizes the ~90 kDa (apparent MW) MDM2 protein.
Recognizes the ~90 kDa (apparent MW) MDM2 protein in A549 cells. May also recognize the ~60 kDa and ~90 kDa isoforms of MDM2.
This Anti-MDM2 (Ab-3) Mouse mAb (4B11) is validated for use in Immunoblotting, Immunofluorescence, Immunoprecipitation, Paraffin Sections for the detection of MDM2 (Ab-3).

Immunogen

Epitope: within amino acids 383-491
Human
full-length, recombinant human MDM2

Application


Immunoblotting (2 g/ml, chemiluminescence)
Immunofluorescence (1 g/ml)
Immunoprecipitation (1 g/reaction)
Paraffin Sections (2.5 g/ml, heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 50% glycerol.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
A549 cells

Other Notes

Marchetti, A., et al. 1995. J. Pathol.175, 31.
Barak, Y., et al. 1993. EMBO. J.12, 461.
Ladanyi, M., et al. 1993. Cancer Res.53, 16.
Leach, F. S., et al. 1993. Cancer Res.53, 2231.
Oliner, J. D., et al. 1993. Nature362, 857.
Momand, J., et al. 1992. Cell69, 1237.
Oliner, J. D., et al. 1992. Nature358, 80.
Fakharzadeh, S. S., et al. 1991. EMBO. J.10, 1565.
May also recognize the ~60 kDa and ~90 kDa isoforms of MDM2. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Kaindl et al.
Leukemia, 28(3), 600-608 (2013-11-19)
ETV6/RUNX1 (E/R) is the most common fusion gene in childhood acute lymphoblastic leukemia. It is responsible for the initiation of leukemia but also indispensable for disease maintenance and propagation, although its function in these latter processes is less clear. We
Junhui Li et al.
Molecular cancer therapeutics, 21(4), 535-545 (2022-02-09)
High frequency of KRAS and TP53 mutations is a unique genetic feature of pancreatic ductal adenocarcinoma (PDAC). TP53 mutation not only renders PDAC resistance to chemotherapies but also drives PDAC invasiveness. Therapies targeting activating mutant KRAS are not available and
Julian Aiken et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(3), 1120-1134 (2015-11-19)
We demonstrated in a previous study that murine double minute (Mdm)-2 is essential for exercise-induced skeletal muscle angiogenesis. In the current study, we investigated the mechanisms that regulate Mdm2 activity in response to acute exercise and identified VEGF-A as a
Lauren E Cowen et al.
Scientific reports, 9(1), 13097-13097 (2019-09-13)
The 14-3-3-related protein SMG7 plays critical roles in regulation of DNA damage response and nonsense-mediated mRNA decay (NMD). Like 14-3-3, SMG7 engages phosphoserine-dependent protein interactions; however, the precise role of phosphorylation-mediated SMG7 binding remains unknown. Here, we show that DNA
Wendy M Swetzig et al.
Oncotarget, 7(13), 16049-16069 (2016-02-26)
MDM2 and MDM4 are heterodimeric, non-redundant oncoproteins that potently inhibit the p53 tumor suppressor protein. MDM2 and MDM4 also enhance the tumorigenicity of breast cancer cells in in vitro and in vivo models and are overexpressed in primary human breast

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service