Skip to Content
Merck
All Photos(3)

Key Documents

MMCABLEMM

Millipore

MILLIFLEX OASIS® electrical cable for connecting two pumps

for use with Milliflex Oasis® systems

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
CHF 98.10

CHF 98.10


Estimated to ship on10 April 2025


Request more information

Select a Size

Change View
1 UNIT
CHF 98.10

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

CHF 98.10


Estimated to ship on10 April 2025


Request more information

packaging

pack of 1 unit

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® systems

shipped in

ambient

General description

MILLIFLEX OASIS® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. The MILLIFLEX OASIS® pump is a new generation vacuum pump for water and bioburden analysis in pharmaceutical environments. The MILLIFLEX OASIS® electrical cable is used for connecting two pumps together and benefit from a high throughput workflow using one power supply. A second cable must be ordered to connect three pumps together.

Application

MILLIFLEX OASIS® filtration pump finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical Products Analysis.

Legal Information

MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service