Skip to Content
Merck
All Photos(1)

Documents

MABX8413

Sigma-Aldrich

Anti-COVID-19, MERS & SARS-CoV, Nucleoprotein Antibody, 1058

suitable for ELISA

Synonym(s):

SARS-CoV-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1058, monoclonal

storage condition

avoid repeated freeze/thaw cycles

technique(s)

ELISA: suitable

isotype

IgG1

shipped in

wet ice

General description

This mouse monoclonal Anti-COVID-19, MERS & SARS-CoV, Nucleoprotein, 1058, Cat. No. MABX8413 is tested for use in ELISA, for the detection of COVID-19 Nucleoprotein.

Specificity

COVID-19, MERS & SARS Coronavirus Nucleoprotein (NP)

Immunogen

Recombinant Coronavirus nucleoprotein peptide

Target description

Coronavirus disease 2019 (COVID-19) is an infectious disease caused by the virus strain severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus closely related to the MERS and SARS viruses.

Physical form

Purified in 10mM Phosphate Buffered Saline with 0.1% sodium azide, pH 7.0.

Reconstitution

Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Short Term: 2-8°C. Long Term: -20°C. Avoid repeated freezing and thawing.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service