Skip to Content
Merck
All Photos(3)

Documents

MABT330

Sigma-Aldrich

Anti-Lamin A (mature) Antibody, clone 4A4-A4

clone 4A4-A4, from mouse

Synonym(s):

Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A4-A4, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LMNA(4000)

General description

Nuclear lamins are composed of the type V intermediate filament proteins. Often referred to as nucleoskeletal proteins they play a key role in nuclear integrity, positioning of nuclear pores, and overall nuclear size and shape. They also play several key functional roles in the maintenance and propagation of the genome--replication, transcription-- as well as the disassembly and reassembly of the nucleus during cell division. In vitro studies have shown that dimers are the basic building blocks of higher order lamin structures and in low concentrations lamins are distributed throughout the nucleoplasm. In humans, there are two types of lamins: A-type lamins (lamins A and C), found primarily in differentiated cells, and B-type lamins (lamins B1 and B2), found in all nucleated cells. Nuclear lamins are involved in a number of essential nuclear functions, including nuclear envelope assembly and disassembly during cell division, DNA synthesis, transcription, and apoptosis. Nuclear lamins have been found to co-localize with DNA synthesis sites.

Immunogen

Epitope: C-terminus
KLH-conjugated Peptide corresponding to the C-terminus of human Lamin A (mature).

Application

Anti-Lamin A (mature), clone 4A4-A4 is an antibody against Lamin A (mature) for use in WB, IP, ICC.
Immunoprecipitation Analysis: A representative lot immunoprecipitated Lamin A (mature) in wild type mouse fibroblasts cell lysate.
Immunocytochemistry Analysis: A representative lot detected Lamin A (mature) in wild type mouse fibroblasts cells.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Lamin A (mature) in 10 µg of A431 and THP-1 cell lysate.
Western Blotting Analysis: A representative lot detected Lamin A (mature) in EDMD and FPLD fibroblasts (Roblek, M., et al. (2010). PLOS ONE. 5(5):e10604).
Western Blotting Analysis: A 1:100 dilution from a representative lot detected Lamin A (mature) in pre-lamin A and Lamin A (mature).
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Lamin A (mature) in 10 µg of Jurkat cell lysate.

Target description

~70 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service