Skip to Content
Merck
All Photos(2)

Key Documents

MABT186

Sigma-Aldrich

Anti-Exo70 Antibody, clone 70X13F3

clone 70X13F3, from mouse

Synonym(s):

Exocyst complex component 7, Exocyst complex component Exo70, rExo70

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 438.00

CHF 438.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
CHF 438.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 438.00


Estimated to ship on09 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

70X13F3, monoclonal

species reactivity

human, rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EXOC7(23265)

General description

The exocyst complex component 70 (Exo70) is a component of the exocyst complex, an octameric structure that is also comprised of Sec3, Sec5, Sec6, Sec8, Sec10, Sec15, Exo70, and Exo84. The exocyst complex facilitates the fusion of secretory vesicles with the plasma membrane and the release of vesicular cargo, which may include neurotransmitters, hormones, and membrane proteins. Previous studies have indicated that Exo70 plays an important role in actin assembly and cell motility.

Immunogen

GST-tagged recombinant protein corresponding to rat Exo70.

Application

Detect Exo70 using this Anti-Exo70 Antibody, clone 70X13F3 validated for use in Western Blotting, ICC.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling
Western Blot Analysis: A 1:1,000 dilution from a representative lot detected Exo70 in 10 µg of human brain tissue lysate.
Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in Exo70 cells. (Hsu, S.C., et al. (2001). The Journal of Neuroscience. 21(11):3839–3848.)
Immunofluorescence Analysis: A previous lot was used by an independent laboratory in IF. (Hsu, S.C., et al. (2011). The Journal of Neuroscience. 31(10):3522–3535.)

Quality

Evaluated by Western Blot in rat brain tissue lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Exo70 in 10 µg of rat brain tissue lysate.

Target description

~75 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilka Wilhelmi et al.
Molecular metabolism, 45, 101151-101151 (2020-12-29)
Hormone secretion from metabolically active tissues, such as pancreatic islets, is governed by specific and highly regulated signaling pathways. Defects in insulin secretion are among the major causes of diabetes. The molecular mechanisms underlying regulated insulin secretion are, however, not
Cristina Cacho-Navas et al.
Cellular and molecular life sciences : CMLS, 79(1), 61-61 (2022-01-10)
Apical localization of Intercellular Adhesion Receptor (ICAM)-1 regulates the adhesion and guidance of leukocytes across polarized epithelial barriers. Here, we investigate the molecular mechanisms that determine ICAM-1 localization into apical membrane domains of polarized hepatic epithelial cells, and their effect
Cristina Cacho-Navas et al.
eLife, 12 (2024-04-10)
Epithelial intercellular adhesion molecule (ICAM)-1 is apically polarized, interacts with, and guides leukocytes across epithelial barriers. Polarized hepatic epithelia organize their apical membrane domain into bile canaliculi and ducts, which are not accessible to circulating immune cells but that nevertheless

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service