Skip to Content
Merck
All Photos(2)

Documents

MABS829

Sigma-Aldrich

Anti-phospho-EGFR (Tyr1173) Antibody, clone 3E7.2

clone 3E7.2, from mouse

Synonym(s):

Epidermal growth factor receptor, EC: 2.7.10.1, Proto-oncogene c-ErbB1, Receptor tyrosine protein kinase erbB-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E7.2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pTyr1173)

Gene Information

human ... EGFR(1956)

General description

Epidermal growth factor receptor (UniProt: P00533; also known as EC: 2.7.10.1; Proto-oncogene c-ErbB1, Receptor tyrosine protein kinase erbB-1) is encoded by the EGFR (also known as ERBB, ERBB1, HER1) gene (Gene ID: 1956) in human. EGFR is activated by binding of its specific ligands and transforms from a monomeric to active homodimeric form. In addition to forming homodimers, EGFR may pair with another member of the ErbB receptor family, such as ErbB2/Her2/neu, to create an activated heterodimer. EGFR dimerization stimulates its intrinsic intracellular protein-tyrosine kinase activity. As a result, autophosphorylation of five tyrosine residues (Y992, Y1045, Y1068, Y1148 and Y1173) occurs in the C-terminal domain. This autophosphorylation elicits downstream activation and signaling by several other proteins that associate with the phosphorylated tyrosines through their own phosphotyrosine-binding domains. EGFR activation results in at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLC gamma-PKC and STATs modules. These downstream events modulate several cell functions, including migration, adhesion, and proliferation. Endocytosis and inhibition of the activated EGFR by phosphatases constitute key regulatory mechanisms.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids surrrounding Tyrosine 1173 of human epidermal growth factor receptor.

Application

Detect phospho EGFR using this mouse monoclonal Anti-phospho-EGFR (Tyr1173), clone 3E7.2, Cat. No. MABS829, used in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: 10 µg/mL from a representative lot detected phospho-EGFR (Tyr1173) in A431 cells treated with EGF.

Quality

Evaluated by Western Blotting in A431 cells treated with EGF.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected phospho-EGFR (Tyr1173) in 10 µg lysate from A431 cells treated with EGF.

Target description

~200 kDa observed; 134.28 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 05-483

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service