Skip to Content
Merck
All Photos(2)

Key Documents

MABS421

Sigma-Aldrich

Anti-DKK3, clone 4G7 Antibody

ascites fluid, clone 4G7, from mouse

Synonym(s):

Dickkopf-related protein 3, Dickkopf-3, Dkk-3, hDkk-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 264.00

CHF 264.00


Estimated to ship on25 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1074

Request a Bulk Order

Select a Size

Change View
100 μL
CHF 264.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 264.00


Estimated to ship on25 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1074

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4G7, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DKK3(27122)

General description

Dickkopf-related protein 3, also known as Dickkopf-3, or Dkk-3, or hDkk-3, and encoded by the gene DKK3/ REIC is a member of the dickkopf family. DKK3 is a 350 amino acid secreted glycoprotein that is composed of an N-terminal signal peptide and two conserved cysteine-rich domains, which are separated by a 12 amino acid linker region. This secreted protein is involved in embryonic development through its interactions with the Wnt signaling pathway. The expression of this gene is decreased in a variety of cancer cell lines and it may function as a tumor suppressor gene.

Immunogen

Purified recombinant fragment of human DKK3 expressed in E. Coli

Application

Anti-DKK3, clone 4G7 Antibody is an antibody against DKK3 for use in western blotting & flow cytometry.
Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected DKK3 in MCF-7 cells.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blotting in HEK293, MCF-7 and HL7702 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected DKK3 in HEK293, MCF-7 and HL7702 cell lysates.

Target description

~44 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service