Skip to Content
Merck
All Photos(1)

Key Documents

MABS275

Sigma-Aldrich

Anti-Gli3 Antibody, clone 5E1

clone 5E1, from mouse

Synonym(s):

Transcriptional activator GLI3, GLI3 form of 190 kDa, GLI3-190, GLI3FL, GLI3 C-terminally truncated form, GLI3 form of 83 kDa, GLI3-83

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 414.00

CHF 414.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
CHF 414.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 414.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E1, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GLI3(2737)

General description

Gli3 can function as both a transcriptional activator and repressor of the sonic hedgehog (Shh) pathway. The full-length Gli3 form (GLI3FL) becomes an activator (GLI3A) after phosphorylation and nuclear translocation. The C-terminally truncated form (GLI3R), acts as a repressor. Proper balance between the Gli3 activator and the repressor specifies limb digit number and identity during development. In concert with TRPS1, Gli3 plays a role in activating chondrocyte proliferation. Gli3 is expressed in several normal adult tissues, including lung, colon, spleen, placenta, testis, and myometrium. Gli3 defects are associated with Greig cephalo-poly-syndactyly syndrome (GCPS), Pallister-Hall syndrome (PHS), polydactyly postaxial type A1 (PAPA1), polydactyly postaxial type B polydactyly (PAPB), and polydactyly preaxial type 4 (POP4).

Specificity

This antibody recognizes the truncated, repressor form and the full length protein of human Gli3.

Immunogen

Recombinant protein corresponding to the putative repressive motif of human GLI3.

Application

Immunocytochemistry Analysis: A representative lot detected Gli3 in PK-15, COS-1, and TM4 cells cotransfected with recombinant Gli3 (Hunt, R., et al. (2007). Hybridoma. 27(4):231-240.).

Immunohistochemistry Analysis: A representative lot detected Gli3 in mouse embryo cross-sections (Laht, S., et al. (2008). Hybridoma. 27(3):167-174.).
Research Category
Signaling
Research Sub Category
Developmental Signaling
This Gli3 antibody is validated for use in WB, ICC & IHC for the detection of the Gli3 protein.

Quality

Evaluated by Western Blotting in human thymus tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected in Gli3 in 10 µg of human thymus tissue lysate.

Target description

~75 kDa and ~200 kDa observed. The truncated, repressor form and full length form of this protein has been observed at ~83 kDa and ~190 kDa, respectively (Tanimoto, Y., et al. (2012). J Biol Chem. 287(25):21429-21438.). Uncharacterized bands may be observed at ~160 kDa and below ~50 kDa in some cell lysates.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human thymus tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Hua Chen et al.
Molecular genetics & genomic medicine, 5(4), 390-404 (2017-07-19)
Alström syndrome (AS), featuring retinal dystrophy, neuronal deafness, cardiomyopathy, metabolic syndrome, and diffuse fibrosis, is caused by biallelic mutations in the centrosomal protein ALMS1. Genotype-phenotype correlation has been suggested without assessment of ALMS1 expression. ALMS1 expression (real-time PCR and immunocytochemistry)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service