Skip to Content
Merck
All Photos(1)

Key Documents

MABS2047M

Sigma-Aldrich

Anti-MT-ND2 Antibody, clone 9E12-1B3

clone 9E12-1B3, from mouse

Synonym(s):

NADH-ubiquinone oxidoreductase chain 2, EC: 1.6.5.3, NADH dehydrogenase subunit 2, MT-ND2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CHF 163.00
100 μG
CHF 372.00

CHF 163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
CHF 163.00
100 μG
CHF 372.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 163.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9E12-1B3, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MT-ND2(4536)

General description

NADH-ubiquinone oxidoreductase chain 2 (UniProt: P03891; also known as EC: 1.6.5.3, NADH dehydrogenase subunit 2, MT-ND2) is encoded by the MT-ND2 (also known as MTND2, NADH2, ND2) gene (Gene ID: 4536) in human. MT-ND2 is a mitochondrial inner membrane protein that forms the core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). It is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (Coenzyme Q10) that participates in the generation of a proton gradient, which is then used for ATP synthesis. Defects in MT-ND2 gene are known to cause of Leber hereditary optic neuropathy (LHON) that is a maternally inherited disease resulting in acute or subacute loss of central vision, due to optic nerve dysfunction. LHON results from primary mitochondrial DNA mutations affecting the respiratory chain complexes.

Specificity

Clone 9E12-1B3 detects NADH-ubiquinone oxidoreductase chain 2 (MT-ND2) in mitochondria from human cells. It targets an epitope within the N-terminal region.

Immunogen

Epitope: N-terminus
Synthetic peptide corresponding to the N-terminus of human MT-ND2.

Application

Anti-MT-ND2, clone 9E12-1B3, Cat. No. MABS2047, is a mouse monoclonal antibody that detects NADH-ubiquinone oxidoreductase chain 2 and has been tested for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: 2 µg/mL from a representative lot detected MT-ND2 in mitochondria from human neonatal dermal fibroblasts and mitochondria from human neonatal dermal fibroblasts depleted of mtDNA (Courtesy of Michael F. Marusich, Ph.D., mAbDx, Inc., Eugene, OR USA).

Quality

Evaluated by Western Blotting in mitochondria from human neonatal dermal fibroblasts and mitochondria from human neonatal dermal fibroblasts depleted of mtDNA.

Western Blotting Analysis: 1 ug/mL of this antibody detected MT-ND2 in 10 µg of human neonatal dermal fibroblasts and mitochondria from human neonatal dermal fibroblasts depleted of mtDNA.

Target description

~30 kDa observed; 38.96 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein L
Purified mouse monoclonal antibody IgG2a in buffer containing HEPES-Buffered Saline (150 mM NaCl, 15 mM HEPES, pH 7.2) with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Sheng Zhang et al.
Nature cell biology, 23(7), 684-691 (2021-07-14)
Members of the mammalian AlkB family are known to mediate nucleic acid demethylation1,2. ALKBH7, a mammalian AlkB homologue, localizes in mitochondria and affects metabolism3, but its function and mechanism of action are unknown. Here we report an approach to site-specifically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service