Skip to Content
Merck
All Photos(1)

Key Documents

MABS1703

Sigma-Aldrich

Anti-MGAT2 Antibody, clone 4A5.2

clone 4A5.2, from mouse

Synonym(s):

Alpha-1, 6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase, Beta-1, 2-N-acetylglucosaminyltransferase II, GlcNAc-T II, GNT-II, Mannoside acetylglucosaminyltransferase 2, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A5.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

glycosylation

Gene Information

human ... MOGAT2(80168)

General description

Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase (EC 2.4.1.143; UniProt Q10469; also known as Beta-1,2-N-acetylglucosaminyltransferase II, GlcNAc-T II, GNT-II, Mannoside acetylglucosaminyltransferase 2, MGAT2, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyl-transferase II) is encoded by the MGAT2 (also known as CDG2A) gene (Gene ID 4247) in human. MGAT2 is a Golgi enzyme that catalyzes an essential step in the conversion of oligo-mannose to complex N-glycans. It possesses the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. It is highly expressed in small intestine in mice and humans. Mutations in MGTA2 gene are known to cause congenital disorders of glycosylation 2A, which is a multi-system disorder caused by defective glycoprotein biosynthesis. It leads to under-glycosylated levels of serum glycoproteins that result in defects in neuronal development, coagulation disorders, hypotonia, and immunodeficiency.

Ref.:
Tan, J., et al. (1995). Eur. J. Biochem. 231, 317-328.
Tan J., et al. (1996), Am. J. Hum. Genet. 159, 810-817.

Specificity

Clone 4A5.2 targets an internal epitope within the C-terminal half of human MGAT2.

Immunogen

GST-tagged internal fragment from the C-terminal half of human MGAT2.

Application

Detect MGAT2 using this mouse monoclonal Anti-MGAT2 Antibody, clone 4A5.2, Cat. No. MABS1703, validated for use in Western Blotting.
Research Category
Signaling

Quality

Evaluated by Western Blotting in T47D cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected MGAT2 in 10 µg of T47D cell lysate.

Target description

~52 kDa observed. 51.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service