Skip to Content
Merck
All Photos(2)

Key Documents

MABS146

Sigma-Aldrich

Anti-RasGRP1 Antibody, clone 10.1

clone 10.1, from mouse

Synonym(s):

RAS guanyl-releasing protein 1, Calcium and DAG-regulated guanine nucleotide exchange factor II, CalDAG-GEFII, Ras guanyl-releasing protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 359.00

CHF 359.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 359.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 359.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RASGRP1(10125)

General description

RasGRP1, also known as Calcium and DAG-regulated guanine nucleotide exchange factor II (CalDAG-GEFII) or Ras guanyl-releasing protein (RASGRP), and encoded by the gene RASGRP1/RASGRP, functions as a diacylglycerol (DAG)-regulated nucleotide exchange factor specifically activating Ras through the exchange of bound GDP for GTP. RasGRP1 activates the Erk/MAP kinase cascade and couples T-lymphocytes and B-lymphocytes antigen receptors to the activation of Ras. Hence, it regulates T-cells and B-cells development, homeostasis and differentiation. Additionally, RasGRP1 functions also in FcERI-evoked degranulation and cytokine secretion by mast cells, regulating allergic responses. RasGRP1 may also function in other cell types’ differentiation. RasGRP1 is localized to the cytoplasm and cell membrane; it is a peripheral membrane protein in the Golgi/ER membranes. In neurons, RasGRP1 localizes to somata and dendrites but not to axons of hippocampal pyramidal cells of in the brain and in particular the cerebellum, cortex and amygdata. RasGRP1 also is expressed in the hematopoietic system including T cells and RasGRP1 is also found expressed developmentally in fetal brain and kidney tissues. Defects in RasGRP1 may be associated with Systemic lupus erythematosus (SLE) a chronic, relapsing, inflammatory, and often febrile multisystemic disorder of connective tissue, characterized principally by involvement of the skin, joints, kidneys and serosal membranes. It is of unknown etiology, but is thought to represent a failure of the regulatory mechanisms of the autoimmune system. The disease is marked by a wide range of system dysfunctions, an elevated erythrocyte sedimentation rate, and the formation of LE cells in the blood or bone marrow. Interestingly it is thought that aberrantly spliced isoforms or other mutations of RASGRP1 give rise to dysregulation of various signaling processes which in turn contribute to the development of autoimmunity in SLE patients.

Immunogen

GST-tagged recombinant protein corresponding to human RasGRP1.

Application

Detect RasGRP1 using this mouse monoclonal antibody, Anti-RasGRP1 Antibody & clone 10.1 validated for use in western blotting.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected RasGRP1 in 10 µg of human cerebellum tissue lysate.
Flow Cytometry Analysis: A representative lot detected RasGRP1 in malignant lymphoid cells treated with tipifarmib (Ding, H., et al. (2011). Blood. 118:4872-4881).

Quality

Evaluated by Western Blotting in MOLT-4 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected RasGRP1 in 10 µg of MOLT-4 cell lysate.

Target description

~90 kDa observed. Uniprot describes 3 isoforms produced by alternative splicing at ~62 kDa, ~55 kDa, and ~68 kDa

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arianna De Rosa et al.
Biomolecules, 12(2) (2022-02-26)
Schizophrenia (SCZ) is a polygenic severe mental illness. Genome-wide association studies (GWAS) have detected genomic variants associated with this psychiatric disorder and pathway analyses have indicated immune system and dopamine signaling as core components of risk in dorsolateral-prefrontal cortex (DLPFC)
Sarah Winter et al.
EMBO molecular medicine, 10(2), 188-199 (2017-12-29)
Inherited CTPS1, CD27, and CD70 deficiencies in humans have revealed key factors of T-lymphocyte expansion, a critical prerequisite for an efficient immunity to Epstein-Barr virus (EBV) infection. RASGRP1 is a T-lymphocyte-specific nucleotide exchange factor known to activate the pathway of
Neelam Shahani et al.
Science signaling, 9(454), ra111-ra111 (2016-12-03)
The striatum of the brain coordinates motor function. Dopamine-related drugs may be therapeutic to patients with striatal neurodegeneration, such as Huntington's disease (HD) and Parkinson's disease (PD), but these drugs have unwanted side effects. In addition to stimulating the release
Mehdi Eshraghi et al.
Science advances, 6(18), eaaz7001-eaaz7001 (2020-05-20)
The therapeutic effects of l-3,4-dihydroxyphenylalanine (l-DOPA) in patients with Parkinson's disease (PD) severely diminishes with the onset of abnormal involuntary movement, l-DOPA-induced dyskinesia (LID). However, the molecular mechanisms that promote LID remain unclear. Here, we demonstrated that RasGRP1 [(guanine nucleotide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service