Skip to Content
Merck
All Photos(1)

Key Documents

MABS1147

Sigma-Aldrich

Anti-SH2D1A Antibody, clone SuGr-20 18A9

clone SuGr-20 18A9, from mouse

Synonym(s):

SH2 domain-containing protein 1A, Duncan disease SH2-protein, Signaling lymphocytic activation molecule-associated protein, SLAM-associated protein, T-cell signal transduction molecule SAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SuGr-20 18A9, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SH2D1A(4068)

General description

SH2 domain-containing protein 1A (UniProt O60880; also known as Duncan disease SH2-protein, Signaling lymphocytic activation molecule-associated protein, SLAM-associated protein, T-cell signal transduction molecule SAP) is encoded by the SH2D1A (also known as DSHP, SAP, XLP, XLP1, XLPD) gene (Gene ID 4068) in human. SAP/SH2D1A is a 628-amino acid (isoform A) adaptor protein expressed in T cells, natural killer (NK) cells, NK-T cells, and some B cells. SAP/SH2D1A is made up almost exclusively of a Src homology 2 (SH2) domain (a.a. 6-104) and plays an important role in multiple immune cell functions, including T helper 2 (TH2) cytokine production, T cell-dependent antibody production, follicular T helper (TFH) cell differentiation, NK-T cell development, CD8+ T cell- and NK cell-mediated cytotoxicity. SAP couples signaling lymphocytic activation molecule (SLAM) family receptors to tyrosine phosphorylation signals through its interaction with the SH3 domain of Src family protein tyrosine kinase Fyn. In addition, SAP can prevent SH2 domain-containing negative regulators, such as phosphatases SHIP-1 and SHP-1, from inactivating SLAM-related receptors by blocking their interaction with the receptors. Evidence also shows that SAP/SH2D1A regulates T cell-dependent antibody production and TFH cell differentiation independent of Fyn. SAP/SH2D1A gene is mutated and inactivated in X-linked lymphoproliferative disease, a rare human immunodeficiency characterized by an uncontrolled immune response to Epstein-Barr virus infection.

Specificity

Clone SuGr-20 18A9 recognizes recombinant full-length human SH2D1A spliced isoform A (UniProt O60880-1) by Western blotting. Reactivity toward spliced isoforms B to F (UniProt O60880-2 to O60880-6) has not been determined.

Immunogen

Recombinant full-length human SH2D1A.

Application

Anti-SH2D1A Antibody, clone SuGr-20 18A9 is an antibody against SH2D1A for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

Evaluated by Western Blotting of recombinant human SH2D1A.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected 0.1 µg of recombinant human SH2D1A.

Target description

~14 kDa observed. 14.19 kDa calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service