Skip to Content
Merck
All Photos(3)

Documents

MABN788

Sigma-Aldrich

Anti-nitrated Hsp90 Antibody, clone 44.23

clone 44.23, from mouse

Synonym(s):

Heat shock protein HSP 90-beta, Heat shock 84 kDa, HSP 84, HSP84

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

44.23, monoclonal

species reactivity

rat

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
dot blot: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Heat shock protein HSP 90-beta (UniProt: P34058; also known as Heat shock 84 kDa, HSP 84, HSP84) is encoded by the Hsp90ab1 (also known as Hsp84, Hspcb) gene (Gene ID: 301252) in rat. HSP90 is a ubiquitous chaperone protein with multiple functions in the cell. It promotes the maturation, structural maintenance and proper regulation of specific target proteins involved for instance in cell cycle control and signal transduction. HSP90 undergoes a functional cycle that is linked to its ATPase activity. This cycle probably induces conformational changes in the client proteins, such as transcription factors, hormone receptors, and protein kinases, thereby causing their activation. HSP90 is abundant protein in the cell and may account for up to 2% of the total cytosolic protein. HSP90 is considered to be absolutely essential for cell survival. Its anti-apoptotic activity involves the inhibition of Apaf-1 oligomerization and caspase activation. Apart from its chaperone activity, HSP90 also plays a role in the regulation of the transcription machinery. HSP90 and its co-chaperones modulate transcription by altering the steady-state levels of certain transcription factors in response to various physiological cues and also modulate the activity of certain epigenetic modifiers, such as histone deacetylases or DNA methyl transferases. HSP90 can also participate in the eviction of histones from the promoter region of certain genes and thereby turn on gene expression. Nitration of HSP90 results in decreased chaperone activity and intracellular release of the nitrated HSP90 is shown to induce induces death of PC12 cells. (Ref.: Ye, Y et al (2007). J. Biol. Chem. 282(9); 6324-6337).

Specificity

Clone 44.23 detects nitrated HSP90 in rat, but not HSP90. It specifically targets nitrotyrosine 56.

Immunogen

Purified, nitrated HSP90 from rat liver.

Application

Anti-Nitrated Hsp90, clone 44.23, Cat. No. MABN788, is a highly specific mouse monoclonal antibody that targets nitrated HSP90 and has been tested in Dot Blot, ELISA, Immunohistochemistry (Paraffin), and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected nitrated HSP90 in Western Blotting applications (Ye, Y., et. al. (2007). J Biol Chem. 282(9):6324-37).

ELISA Analysis: A representative lot detected nitrated HSP90 in ELISA applications (Ye, Y., et. al. (2007). J Biol Chem. 282(9):6324-37).

Dot Blot Analysis: A representative lot detected nitrated HSP90 in Dot Blot applications (Franco, M.C., et. al. (2013). Proc Natl Acad Sci U S A. 110(12):E1102-11).

Immunohistochemistry Analysis: A representative lot detected nitrated HSP90 in Immunohistochemistry applications (Franco, M.C., et. al. (2013). Proc Natl Acad Sci U S A. 110(12):E1102-11).

Quality

Evaluated by Western Blotting in PC12 cells treated with pyroxynitrite.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected nitrated HSP90 in PC12 cells treated with pyroxynitrite.

Target description

90 kDa observed; 83.28 kda calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service