Skip to Content
Merck
All Photos(2)

Key Documents

MABN726

Sigma-Aldrich

Anti-EphB2 Antibody, clone 2D12C6

ascites fluid, clone 2D12C6, from mouse

Synonym(s):

Ephrin type-B receptor 2, EPHB2 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 424.00

CHF 424.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
CHF 424.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 424.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2D12C6, monoclonal

species reactivity

rat, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Ephrin type-B receptor 2 (EphB2) is a receptor tyrosine kinase for ephrin ligands that mediate numerous developmental processes, particularly in the nervous system. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB2 is critical for embryonic/postnatal development for lateral suprapyramidal blade formation which is in a specific region of the dentate gyrus. EphB2 is expressed in the two main components of the thymus, developing thymocytes, TEC, colorectal cancer, and in the earliest SGZ progenitor cells as they migrate during development and form the dentate gyrus.

Immunogen

Purified recombinant fragment of EphB2 expressed in E. coli.

Application

Anti-EphB2 Antibody, clone 2D12C6 is a highly specific mouse monoclonal antibody, that targets EphB2 & has been tested in western blotting, IHC & ICC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected EphB2 in HeLa and HepG2 cells.

Immunohistochemistry Analysis: 1:200 to 1:1,000 dilution from a representative lot detected EphB2 in rat cerebellum, pons/midbrain, and hippocampus tissues. Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in IMR-32 cell lysate.

Western Blot Analysis: A 1:500 dilution of this antibody detected EphB2 in 10 µg of IMR-32 cell lysate.

Target description

~53 kDa observed

Physical form

Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
IMR-32 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service