Skip to Content
Merck
All Photos(1)

Key Documents

MABN52

Sigma-Aldrich

Anti-REP-1 Antibody, clone 2F1

clone 2F1, from mouse

Synonym(s):

choroideremia (Rab escort protein 1), choroideremia, Choroideraemia protein, Rab escort protein 1, Rab geranylgeranyltransferase component A, TCD protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHM(1121)

General description

Rab proteins are low molecular weight, ras-related GTPases that bind to a cell membrane’s cytoplasmic surface and function to regulate protein trafficking in both endocytic and secretory pathways. Rab escort protein (REP)-1 is a 653 amino acid protein belonging to the Rab GDI family of proteins. REP-1 binds to newly assembled unprenylated RAB proteins, escorts them to the Rab GGTases, and remains attached during and after the geranylgeranyl transfer reaction. REP proteins are composed of two conserved domains connected by a 150 amino acid insert. The multisheet domain I acts to assemble a Rab-binding platform. A globular, smaller, α-helical domain II, is thought to participate in membrane-protein interaction. Defects in the expression of REP-1 cause truncation or absence of the protein, and result in a disease called choroideremia (CHM), an X-linked blindness characterized by a progressive dystrophy of the retinal pigment epithelium, retina, and the choroid.

Specificity

This antibody recognizes REP-1.

Immunogen

Recombinant protein corresponding to human REP-1.

Application

Anti-REP-1 Antibody, clone 2F1 is an antibody against REP-1 for use in WB, IP, IC, IH.
Immunohistochemistry Analysis: A previous lot of MABN52 was successfully used in IH, as reported by an independent laboratory (MacDonald, I.M., et al. (2005). Invest Ophthalmol Vis Sci. 46:E-Abstract 540).

Quality

Evaluated by Western Blot in 293T cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected REP-1 on 10 µg of 293T cell lysate.

Target description

~83 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lewis E Fry et al.
Translational vision science & technology, 10(8), 12-12 (2021-07-14)
Choroideremia results from the deficiency of Rab Escort Protein 1 (REP1), encoded by CHM, involved in the prenylation of Rab GTPases. Here, we investigate whether the transcription and expression of other genes involved in the prenylation of Rab proteins correlates
Nicolas Cereso et al.
Molecular therapy. Methods & clinical development, 1, 14011-14011 (2014-01-01)
Inherited retinal dystrophies (IRDs) comprise a large group of genetically and clinically heterogeneous diseases that lead to progressive vision loss, for which a paucity of disease-mimicking animal models renders preclinical studies difficult. We sought to develop pertinent human cellular IRD
Maria I Patrício et al.
Molecular therapy. Methods & clinical development, 17, 99-106 (2020-01-01)
Recent advances in recombinant adeno-associated virus (rAAV) gene therapy for choroideremia show gene replacement to be a promising approach. It is, however, well known that contact of vector solution with plastic materials in the surgical device may result in non-specific
Nejla Erkilic et al.
Cells, 8(9) (2019-09-14)
Induced pluripotent stem cells (iPSCs) have revolutionized the study of human diseases as they can renew indefinitely, undergo multi-lineage differentiation, and generate disease-specific models. However, the difficulty of working with iPSCs is that they are prone to genetic instability. Furthermore
Maria I Patrício et al.
Molecular therapy. Methods & clinical development, 9, 288-295 (2018-05-01)
Choroideremia (CHM) is a rare, X-linked recessive retinal dystrophy caused by mutations in the CHM gene. CHM is ubiquitously expressed in human cells and encodes Rab escort protein 1 (REP1). REP1 plays a key role in intracellular trafficking through the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service