Skip to Content
Merck
All Photos(2)

Key Documents

MABN502

Sigma-Aldrich

Anti-SNAT1 Antibody, clone N104/37

clone N104/37, from mouse

Synonym(s):

Sodium-coupled neutral amino acid transporter 1, Amino acid transporter A1, rATA1, Glutamine transporter, N-system amino acid transporter 2, Solute carrier family 38 member 1, System A amino acid transporter 1, System A transporter 2, System N amino acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

N104/37, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC38A1(81539)

General description

Sodium-coupled neutral amino acid transporter 1 (SNAT) is also called, Amino acid transporter A1 (ATA1), Glutamine transporter (Glnt), N-system amino acid transporter 2, Solute carrier family 38 member 1 (SLC38A1), System A amino acid transporter 1 (SAT1), System A transporter 2 (SA2), and System N amino acid transporter 1. SNAT functions as a sodium-dependent amino acid transporter that may supply glutamatergic and GABAergic neurons with the glutamine required for the synthesis of the neurotransmitters glutamate and GABA. SNAT is specifically expressed in brain, with the highest levels in cerebellum and thalamus, in glutamatergic, GABAergic and dopaminergic neurons, ependymal cells lining the ventricle and is also detected in spinal cord, heart, colon and placenta.

Specificity

This antibody recognizes the N-terminus of SNAT1.

Immunogen

Epitope: N-terminus
Recombinant protein corresponding to the N-terminus of rat SNAT1.

Application

Detect Sodium-coupled neutral amino acid transporter 1 using this mouse monoclonal antibody, Anti-SNAT1 Antibody, clone N104/37 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected SNAT1 in rat midbrain and human thalamus tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected SNAT1 in 10 µg of human brain tissue lysate.

Target description

~ 54 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanthie Thamotharan et al.
PloS one, 12(5), e0176493-e0176493 (2017-05-04)
Placental insufficiency leading to intrauterine growth restriction (IUGR) demonstrates perturbed gene expression affecting placental angiogenesis and nutrient transfer from mother to fetus. To understand the post-transcriptional mechanisms underlying such placental gene expression changes, our objective was to identify key non-coding
Karen J Gibbins et al.
Biology of reproduction, 98(5), 695-704 (2018-01-20)
Hypertensive disease of pregnancy (HDP) with placental insufficiency is the most common cause of fetal growth restriction (FGR) in the developed world. Despite the known negative consequences of HDP both to the mother and fetus, little is known about the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service