Skip to Content
Merck
All Photos(2)

Key Documents

MABN250

Sigma-Aldrich

Anti-RA-induced protein 17 Antibody, clone 5H5.1

clone 5H5.1, from mouse

Synonym(s):

Retinoic acid-induced protein 17, ZMIZ1, Zinc finger MIZ domain-containing protein 1, PIAS-like protein Zimp10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 334.00

CHF 334.00


Estimated to ship on05 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 334.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 334.00


Estimated to ship on05 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H5.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZMIZ1(57178)

General description

Retinoic acid-induced protein 17 (RAI17) is also called Zinc finger MIZ domain-containing protein 1 (ZMIZ1), and PIAS-like protein Zimp10. Retinoic acid-induced protein 17 promotes AR sumoylation and increases AR ligand-dependent transcriptional activity. Retinoic acid-induced protein 17 is highly expressed in ovary and at lower levels in prostate, spleen and testis.

Immunogen

Recombinant protein corresponding to human RA-induced protein 17.

Application

Detect Retinoic acid-induced protein 17 using this mouse monoclonal antibody, Anti-RA-induced protein 17 Antibody, clone 5H5.1 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected RA-induced protein 17 in human thalamus tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected RA-induced protein 17 in 10 µg of SH-SY5Y cell lysate.

Target description

~115 kDa observed. Uniprot describes 2 isoforms produced by alternative splicing at ~115 kDa and ~107 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sameer Aryal et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(18) (2021-04-29)
Loss of the fragile X mental retardation protein (FMRP) causes fragile X syndrome (FXS). FMRP is widely thought to repress protein synthesis, but its translational targets and modes of control remain in dispute. We previously showed that genetic removal of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service