Skip to Content
Merck
All Photos(1)

Documents

MABN2456

Sigma-Aldrich

Anti-HAP1 Antibody, clone 1B6

clone 1B6, from mouse

Synonym(s):

Huntingtin-associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B6, monoclonal

species reactivity

rat, mouse

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

rat ... Hap1(29430)

General description

Huntingtin-associated protein 1 (UniProt: P54256; also known as HAP1) is encoded by the Hap1 gene (Gene ID: 29430) in rat. HAP1 is a huntingtin-associated protein that is enriched in the brain. It specifically associates with HTT/huntingtin and this binding is enhanced by an expanded polyglutamine repeats within HTT that affects HAP1 interaction properties. Both HTT and HAP1 are reported to be involved in intracellular trafficking and HAP1 is proposed to link HTT to motor proteins and/or transport cargos. HAP1 localizes to large nonmembrane-bound cytoplasmic bodies (stigmoid bodies) in various types of neurons. The formation of stigmoid bodies is regulated by the ratio of isoform A to isoform B of HAP1. HAP1 plays a role in vesicular transport within neurons that involves microtubule-associated transporters and is shown to be attenuated by association with mutant HTT. In rodents, two HAP1 isoforms (HAP1A and HAP1B) have been described that differ in their carboxy terminal. However, humans contain only the HAP1A isoform. In murine models, targeted disruption of the HAP1 gene results in significantly depressed feeding behavior and premature postnatal death. Pups with Hap1 -/- show decreased feeding, malnutrition, dehydration, and premature death. (Ref.: Tang, TS et al., (2003). Neuron 39 (2); 227-239; Chan, EY., et al. (2002). Hum. Mol. Gen. 11(8); 945-959).

Specificity

Clone 1B6 detects both HAP1A and HAP1B proteins in mouse and rat brain. It targets an epitope from the internal region.

Immunogen

Epitope: cytoplasmic domain
GST-tagged recombinant fragment corresponding to 137 amino acids from the internal region of rat Huntingtin-associated protein 1 (HAP1).

Application

Anti-HAP1, clone 1B6, Cat. No. MABN2456, is a highly specific mouse monoclonal antibody that targets Huntingtin-associated protein 1 and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected HAP1 in Immunocytochemistry applications (Fujinaga, R., et. al. (2011). Exp. Cell Res. 317(12):1689-700).

Immunoprecipitation Analysis: A representative lot immunoprecipitated HAP1 in Immunoprecipitation applications (Fujinaga, R., et. al. (2011). Exp. Cell Res. 317(12):1689-700).

Western Blotting Analysis: A representative lot detected HAP1 in Western Blotting applications (Chan, E.Y., et. al (2002). Hum Mol Genet. 11(8):945-59; Tang, T.S., et. al. (2003). Neuron. 39(2):227-39; Torre, E.R., et. al. (2003). J Neurosci Methods. 125(1-2):27-32).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in brain tissue lysate from P1 Balb/c mouse.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HAP1 in brain tissue lysate from day one post-natal (P1) Balb/c mouse.

Target description

~90 kDa observed; 70.21 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service