Skip to Content
Merck
All Photos(1)

Key Documents

MABN2426M

Sigma-Aldrich

Anti-TrpV2 Antibody, clone 2D6

clone 2D6, from mouse

Synonym(s):

Transient receptor potential cation channel subfamily V member 2, Osm-9-like TRP channel 2, OTRPC2, Stretch-activated channel 2B, Vanilloid receptor-like protein 1, VRL-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CHF 163.00
100 μL
CHF 352.00

CHF 163.00


Estimated to ship on05 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
CHF 163.00
100 μL
CHF 352.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

CHF 163.00


Estimated to ship on05 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D6, monoclonal

species reactivity

mouse, hamster, rat

packaging

antibody small pack of 25 μL

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Trpv2(22368)
rat ... Trpv2(29465)

General description

Transient receptor potential cation channel subfamily V member 2 (UniProt: Q9WUD2; also known as TrpV2, Osm-9-like TRP channel 2, OTRPC2, Stretch-activated channel 2B, Vanilloid receptor-like protein 1, VRL-1) is encoded by the Trpv2 (also known as Sac2b, Vrl1) gene (Gene ID: 29465) in murine species. TrpV2 is an ubiquitously expressed, calcium-permeable, non-selective cation channel with an outward rectification. It has been implicated in various physiological and pathological processes, including growth factor signaling, macrophage migration, cancer cell metastasis, heart disease, and neuronal cell development. It appears to be regulated by growth factors, like IGF1, PDGF, and morphogenetic neuropeptide/head activator. It is also activated by temperatures higher than 52C, but is not activated by vanilloids and acidic pH. TrpV2 channel is upregulated by the nerve growth factor (NGF) via the mitogen-activated protein kinase (MAPK) signaling pathway to augment neurite outgrowth. Although NGF increases TrpV2 content, it does not induce the translocation of TrpV2 from cytoplasm to cell surface in PC12 cells. (Ref.: Cohen MR et al. (2015). Mol. Cell Biol. 35(24): 4238-4252).

Specificity

Clone 2D6 detects Transient receptor potential cation channel subfamily V member 2 (TrpV2) in Hamster, Mouse, and Rat.

Immunogen

A full length recombinant rat Transient receptor potential cation channel subfamily V member 2 (TrpV2).

Application

Anti-TrpV2, clone 2D6 Antibody, Cat. No. MABN2426, is a highly specific mouse monoclonal antibody that targets TRPV2 and has been tested in Immunoprecipitation and Western Blotting.
Western Blotting Analysis: A reperesentative lot detected TrpV2 in Western Blotting applications (Cohen, M.R., et. al. (2013). PLoS One. 8(12):e85392; Cohen, M.R., et. al. (2015). Mol Cell Biol. 35(24):4238-52).

Immunoprecipitation Analysis: A reperesentative lot detected TrpV2 in Immunoprecipitation applications (Cohen, M.R., et. al. (2013). PLoS One. 8(12):e85392).

Quality

Evaluated by Western Blotting in PC12 cells treated with NGF.

Western Blotting Analysis: A 1:500 dilution of this antibody detected TrpV2 in 10 µg of PC12 cells treated with NGF 7S (100 ng/72 hours).

Target description

~86 kDa observed; 86.71 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service