Skip to Content
Merck
All Photos(2)

Key Documents

MABN2265

Sigma-Aldrich

Anti-CHD-8 Antibody, clone 7F4.1

clone 7F4.1, from mouse

Synonym(s):

Chromodomain-helicase-DNA-binding protein 8, EC: 3.6.4.12, ATP-dependent helicase CHD8, Helicase with SNF2 domain 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F4.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CHD8(57680)

General description

Chromodomain-helicase-DNA-binding protein 8 (UniProt: Q9HCK8; also known as EC: 3.6.4.12, CHD-8, ATP-dependent helicase CHD8, Helicase with SNF2 domain 1) is encoded by the CHD8 (also known as HELSNF1, KIAA1564) gene (Gene ID: 57680) in human. CHD-8 is a member of the SNF2/RAD54 helicase family. Two isoforms of CHD-8 have been reported that are produced by alternative splicing. It is a DNA helicase that acts as a chromatin remodeling factor and regulates transcription. It also acts as a transcription repressor by remodeling chromatin structure and recruiting histone H1 to target genes. CHD-8 is reported to suppress p53/TP53-mediated apoptosis by recruiting histone H1 and preventing p53/TP53 transactivation activity and also acts as a negative regulator of Wnt signaling pathway by regulating beta-catenin activity. It negatively regulates beta-catenin-targeted gene expression by being recruited specifically to the promoter regions of several beta-catenin responsive genes. CHD-8 is involved in both enhancer blocking and epigenetic remodeling at chromatin boundary via its interaction with CTCF. Mutations in CHD8 gene are associated with Autism 18 that is characterized by impairments in reciprocal social interaction and communication and developmental abnormalities.

Specificity

Clone 7F4.1 detects human Chromodomain-helicase-DNA-binding protein 8 (CHD8). It targets an epitope within 112 amino acids from the C-terminal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 112 amino acids rom the C-terminal half of human Chromodomain-helicase-DNA-binding protein 8. The immunogen sequence is conserved in both isoforms.

Application

Anti-CHD-8, clone 7F4.1, Cat. No. MABN2265, is a highly specific mouse monoclonal antibody that targets Chromodomain-helicase-DNA-binding protein 8 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected CHD-8 in human cervical cancer and human tonsil tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected CHD-8 in 10 µg of SH-SY5Y cell lysate.

Target description

270 and 300 kDa observed; 290.52 and 262.35 kDa calculated for isoforms 1 and 2, respectively. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service