Skip to Content
Merck
All Photos(2)

Documents

MABN1806

Sigma-Aldrich

Anti-TPP1 Antibody, clone 2E12

clone 2E12, from mouse

Synonym(s):

Tripeptidyl-peptidase 1, Cell growth-inhibiting gene 1 protein, Lysosomal pepstatin-insensitive protease, LPIC, Tripeptidyl aminopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E12, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TPP1(1200)

General description

Tripeptidyl-peptidase 1 (UniProt: O14773; also known as TPP1, Cell growth-inhibiting gene 1 protein, Lysosomal pepstatin-insensitive protease, LPIC, Tripeptidyl aminopeptidase) is encoded by the TPP1 (also known as CLN2, GIG1, UNQ267/PRO304) gene (Gene ID: 1200) in human. TPP1 is a lysosomal serine protease that acts as a non-specific peptidase and sequentially removes tripeptides from the free N termini of small polypeptides produced by lysosomal proteinases. TPP1 is expressed in several tissues, with tissues associated with peptide hormone production displaying higher level of expression. Human TPP I is synthesized as a zymogen with an apparent mass of 68 kDa, which is processed to a 48 kDa mature enzyme by removal of 179 amino acid prodomain. TPP I zymogen has five potential N-glycosylation sites at amino acid positions 210, 222, 286, 313, and 443. The N-linked glycans can add around 10 kDa to the molecular mass of TPP 1. Like many other lysosomal hydrolases, TPP I proenzyme is able to autoactivate in the acidic pH in vitro. Mutations in TPP1 gene are shown to be a causative factor in ceroid lipofuscinosis, neuronal, 2 (CLN2) a progressive neurodegenerative, lysosomal storage disease, characterized by intracellular accumulation of autofluorescent liposomal material and clinically manifested seizures, dementia, visual loss, and/or cerebral atrophy.

Specificity

Clone 2E12 specifically immunostains tripeptidyl peptidase 1 in human placenta, heart, and pancreas.

Immunogen

A recombinant fragment corresponding to amino acids 195-563 of human TPP1.

Application

Anti-TPP1, clone 2E12, Cat. No. MABN1806, is a highly specific mouse monoclonal antibody, that targets TPP1/Tripeptidyl-peptidase 1 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:5,000 dilution from a representative lot detected TPP1 in human heart and human pancreas tissue.

Western Blotting Analysis: A 1:50 dilution from a representative lot detected TPP1 in CHO cells transfected with TPP1 and TPP1 mutant (Courtesy of Adam Golabek, Ph.D., New York State Institute for Basic Research in Developmental Disabilities, New York, USA).

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected TPP1 in CHO cells expressing WT TPP1 (Courtesy of Adam Golabek, Ph.D., New York State Institute for Basic Research in Developmental Disabilities, New York, USA).




Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry in human placenta tissue.

Immunohistochemistry Analysis: A 1:5,000 dilution of this antibody detected TPP1 in human placenta tissue.

Target description

61.25 kDa calculated (composed of 6 identical subunits)

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service