MABN1768
Anti-TTBK1 Antibody, clone F287-1.1-1E9
clone F287-1.1-1E9, from mouse
Synonym(s):
Tau-tubulin kinase 1, Brain-derived tau kinase
About This Item
Recommended Products
biological source
mouse
antibody form
purified antibody
antibody product type
primary antibodies
clone
F287-1.1-1E9, monoclonal
species reactivity
mouse, human
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... TTBK1 (84630)
General description
Specificity
Immunogen
Application
Immunohistochemistry Analysis: A representative lot detected a much higher TTBK1 immunoreactivity in the subiculum and external pyramidal layer of visual cortex in paraformaldehyde-fixed frozen brain sections from transgenic mice overexpressing human TTBK1 (Sato, S., et al. (2008). J. Neurosci. 28(53):14511-14521).
Western Blotting Analysis: Representative lots detected both the full-length and the proteolytically processed murine TTBK1 in tissues (cerebellum, cortex, spinal cord) from wild-type mice, as well as the expression of human TTBK1 transgene in tissues from transgenic mice (Sato, S., et al. (Xu, J., et al. (2010). FASEB J. 24(8):2904-2915; 2008). J. Neurosci. 28(53):14511-14521).
Western Blotting Analysis: A representative lot detected an upregulation of both the full-length and the proteolytically processed TTBK1 in the frontal cortex gray matter extracts from Alzheimer′s diseased (AD) human brains than age-matched control non-AD brains (Sato, S., et al. (2008). J. Neurosci. 28(53):14511-14521).
Neuroscience
Neurodegenerative Diseases
Quality
Western Blotting Analysis: 2.5 µg/mL of this antibody detected the expression of human TTBK1 transgene in 20 µg of transgenic mouse brain tissue lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service