MABN1188
Anti-Tyrosine Hydroxylase Antibody, clone 10F8.2
clone 10F8.2, from mouse
Synonym(s):
TH, Tyrosine 3-monooxygenase, Tyrosine 3-hydroxylase, Tyrosine Hydroxylase
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
10F8.2, monoclonal
species reactivity
human, rat
species reactivity (predicted by homology)
mouse (based on 100% sequence homology)
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG2bλ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... TH (7054)
General description
Specificity
Immunogen
Application
Neuroscience
Neuronal & Glial Markers
Quality
Western Blotting Analysis: 0.25 µg/mL of this antibody detected tyrosine hydroxylase in 10 µg of PC12 cell lysate.
Target description
Linkage
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service