Skip to Content
Merck
All Photos(1)

Key Documents

MABF2237

Sigma-Aldrich

Anti-NKp30 (CD337) Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CHF 154.00
100 μL
CHF 391.00

CHF 154.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
CHF 154.00
100 μL
CHF 391.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 154.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C01, monoclonal

mol wt

calculated mol wt 24.93 kDa
observed mol wt ~32 kDa

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

activity assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... NCR3(259197)

General description

Natural cytotoxicity triggering receptor 3 (UniProt: O14931; also known as Activating natural killer receptor p30, Natural killer cell p30-related protein, NK-p30, NKp30, CD337) is encoded by the NCR3 (also known as 1C7, LY117) gene (Gene ID: 259197) in human. NKp30 is a single-pass type I membrane protein that is selectively expressed by all resting and activated NK cells. Its weaker expression is observed in spleen. It serves as a receptor of natural killer/NK cell. It is activated by binding of extracellular ligands including BAG6 and NCR3LG1. It stimulates NK cells cytotoxicity toward neighboring cells producing these ligands. It cooperates with NKp46 and/or NKp44 in inducing NK-mediated cytotoxicity against the majority of target cells. It is also reported to control NK cells cytotoxicity against tumor cells. Its interaction with BAG6 promotes myeloid dendritic cells (DC) maturation, both through killing DCs that fail to achieve a mature phenotype and by inducing the release of TNF-alpha and interferon-gamma by NK cells that promote DC maturation. NKp30 is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 19-135), a transmembrane domain (aa 136-156), and a cytoplasmic domain (aa 157-201). Six isoforms of NKp30 have been described that are produced by alternative splicing.

Specificity

Clone 1C01 is a mouse monoclonal antibody that detects human Natural cytotoxicity triggering receptor 3 (NKp30).

Immunogen

YT cells

Application

Quality Control Testing

Evaluated by Western Blotting in Human spleen tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected NKp30 (CD337) in Human spleen tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected NKp30 (CD337) in Human tonsil tissue lysate.

Function Analysis: A representative lot of this antibody blocked the binding of NKp30-Fc to β-1,3-Glucan and reduced Nkp30 clustering. (Li, S.S., et. al. (2018). Nat Commun. 9(1):751; Li, S.S., et. al. (2013). Cell Host Microbe. 14(4):387-97).

Western Blotting Analysis: A representative lot detected NKp30 (CD337) in Western Blotting applications (Li, S.S., et. al. (2018). Nat Commun. 9(1):751; Li, S.S., et. al. (2013). Cell Host Microbe. 14(4):387-97).

Immunoprecipitation Analysis: A representative lot immunoprecipitated NKp30 (CD337) in Immunoprecipitation applications ( Li, S.S., et. al. (2013). Cell Host Microbe. 14(4):387-97 ).

Flow Cytometry Analysis: A representative lot detected NKp30 (CD337) in Flow Cytometry applications (Li, S.S., et. al. (2018). Nat Commun. 9(1):751; Li, S.S., et. al. (2013). Cell Host Microbe. 14(4):387-97).

Immunocytochemistry Analysis: A representative lot detected NKp30 (CD337) in Immunocytochemitry applications (Li, S.S., et. al. (2013). Cell Host Microbe. 14(4):387-97).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-NKp30 (CD337), clone 1C01, Cat. No. MABF2237, is a This mouse monoclonal antibody that detects NKp30 (Cd337) and is tested for use in Western Blotting, Flow Cytometry, Function Analysis, Immunocytochemistry, and Inhibits Acitivty/Function.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service