Skip to Content
Merck
All Photos(1)

Key Documents

MABF2047

Sigma-Aldrich

Anti-BST-2 (CD317) Antibody, clone 927

clone 927, from rat

Synonym(s):

Bone marrow stromal antigen 2, BST-2, HM1.24 antigen, CD317

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CHF 139.00
100 μG
CHF 341.00

CHF 139.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
CHF 139.00
100 μG
CHF 341.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 139.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

927, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
neutralization: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Bst2(69550)

Related Categories

General description

Bone marrow stromal antigen 2 (UniProt: Q8R2Q8; also known as BST-2, HM1.24 antigen, PDCA-1, CD317) is encoded by the Bst2 gene (Gene ID: 69550) in murine species. BST-2 is a single-pass type II disulfide-linked homodimeric membrane protein that shuttles between the cell membrane, where it is present predominantly in membrane/lipid rafts, and the trans-Golgi network. Its dimerization is essential for its antiviral activity. BST-2 is also shown to form a complex with MMP-14 and localize to the cytoplasm. BST-2 is highly expressed on type I interferon-producing cells in naïve mice. Its levels in most cells are elevated following treatment with type I IFNs and IFN-gamma. It serves as an IFN-induced antiviral host restriction factor that efficiently blocks the release of diverse mammalian enveloped viruses by directly tethering nascent virions to the membranes of infected cells. It acts as a direct physical tether, holding virions to the cell membrane and linking virions to each other. The tethered virions can be either internalized by endocytosis and subsequently degraded or they can remain on the cell surface. In either case, their spread as cell-free virions is restricted. BST-2 is synthesized with a propeptide (aa 153-172), which is removed to generate the mature form. The mature form has a cytoplasmic domain (aa 1-30), a transmembrane domain (aa 31-51), and the extracellular domain (aa 52-152). Within the extracellular region, it has a coiled coil domain (aa 74-147) that forms an extended 17 nm long semi-flexible rod-like structure, which is essential for virion retention at the cell surface and prevent virus spreading.

Specificity

Clone 927 is a rat monoclonal antibody that specifically detects murine Bone marrow stromal antigen 2 (BST-2). It targets an epitope with in the extracellular domain.

Immunogen

Type I IFN-producing cells derived from mouse bone marrow.

Application

Anti-BST-2 (CD317), clone 927, Cat. No. MABF2047, is a rat monoclonal antibody that detects Bone marrow stromal antigen 2 and has been tested for use in ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), and Neutralizing activity.
Neutralizing Analysis: A representative lot efficiently depleted type I interferon-producing cells (IPC) and abrogated IFN-alpha secretion by IPC in response to CpG. (Blasius, A.L., et. al. (2006). J Immunol. 177(5):3260-5).

Immunohistochemistry (Paraffin) Analysis: A representative lot detected BST-2 (CD317) in Immunohistochemistry applications (Yun, T., et. al. (2016). Cell Metab. 23(5):852-66).

ELISA Analysis: A representative lot detected BST-2 (CD317) in ELISA applications (Blasius, A.L., et. al. (2006). J Immunol. 177(5):3260-5).

Flow Cytometry Analysis: A representative lot detected BST-2 (CD317) in Flow Cytometry applications (Blasius, A.L., et. al. (2006). J Immunol. 177(5):3260-5; Yun, T., et. al. (2016). Cell Metab. 23(5):852-66).

Quality

Evaluated by Immunohistochemistry (Paraffin) in mouse spleen and mouse kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:25 dilution of this antibody detected BST-2 (CD317) in mouse spleen and mouse kidney tissue sections.

Target description

~19.15 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service