Skip to Content
Merck
All Photos(1)

Key Documents

MABF174A

Sigma-Aldrich

Anti-CD3 (human), violetFluor® 450, clone SK7 Antibody

clone SK7, from mouse, violetFluor® 450

Synonym(s):

T-cell surface glycoprotein CD3 epsilon chain, T-cell surface antigen T3/Leu-4 epsilon chain, CD antigen CD3e, CD3

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
CHF 188.00
100 TESTS
CHF 405.00

CHF 188.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
25 TESTS
CHF 188.00
100 TESTS
CHF 405.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 188.00


Estimated to ship on13 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

violetFluor® 450

antibody form

purified antibody

antibody product type

primary antibodies

clone

SK7, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CD3E(916)

Application

Anti-CD3 (human), violetFluor® 450, clone SK7 Antibody is an antibody against CD3 for use in Flow Cytometry.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 1 μg from a representative lot detected CD3 in one million human peripheral blood lymphocytes.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

violetFluor is a registered trademark of Tonbo Biotechnologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service