Skip to Content
Merck
All Photos(1)

Key Documents

MABE945

Sigma-Aldrich

Anti-E3 ubiquitin-protein ligase UHRF1 Antibody, clone 7C8

clone 7C8, 1 mg/mL, from mouse

Synonym(s):

E3 ubiquitin-protein ligase UHRF1, Inverted CCAAT box-binding protein of 90 kDa, Nuclear protein 95, Nuclear zinc finger protein Np95, HuNp95, hNp95, RING finger protein 106, Transcription factor ICBP90, Ubiquitin-like PHD and RING finger domain-containi

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
CHF 401.00

CHF 401.00


Estimated to ship on30 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB2114

Request a Bulk Order

Select a Size

Change View
200 μG
CHF 401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 401.00


Estimated to ship on30 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB2114

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7C8, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UHRF1(29128)

General description

Ubiquitin-like with PHD and ring finger domains 1 (UHRF1), also known as ICBP90 or Np95, is a multifunctional nuclear protein that may be an important mediator of heterochromatin formation which is indicative of gene repression. UHRF1 contains a set and RING-finger-associated domain that detects hemi-methylated cytosine residues leading to the recruitment of DNMT1 to these residues during the S phase. UHRF1 thereby facilitates the transfer of methylation status to newly synthesized DNA molecules. UHRF1 may also interact with histones and HDAC1 via its PHD domain to promote modifications of histone residues which contribute to heterochromatin formation. UHRF1 may also directly modify histone H3 by ubiquitination. In addition, UHRF1 may contribute to the DNA-repair process though associations with PCNA. Several studies have suggested that overexpression of UHRF1 may contribute to the development of many cancers.

Immunogen

Recombinant protein corresponding to human E3 ubiquitin-protein ligase UHRF1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-E3 ubiquitin-protein ligase UHRF1 Antibody, clone 7C8 is validated for use in Immunocytochemistry and Chromatin Immunoprecipitation (ChIP) and Western Blotting for the detection of E3 ubiquitin-protein ligase UHRF1.
Western Blotting Analysis: A representative lot detected E3 ubiquitin-protein ligase UHRF1 in overexpressed lysate (Martin Walsh, Mount Sinai School of Medicine).
Chromatin Immunoprecipitation Analysis: A representative lot immunoprecipitated E3 ubiquitin-protein ligase UHRF1 in NTera2 cells (Martin Walsh, Mount Sinai School of Medicine).

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:50 dilution of this antibody detected E3 ubiquitin-protein ligase UHRF1 in HeLa cells.
Alexa Fluor is a registered trademark of Life Technologies.

Target description

90 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service