Skip to Content
Merck
All Photos(2)

Documents

MABE662

Sigma-Aldrich

Anti-TERT Antibody, clone 7D5.2

clone 7D5.2, from mouse

Synonym(s):

Telomerase reverse transcriptase, hEST2, hTRT, Telomerase-associated protein 2, Telomerase catalytic subunit, TP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7D5.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TERT(7015)

General description

Telomerase reverse transcriptase (EC 2.7.7.49; UniProt O14746; also known as hEST2, hTRT, Telomerase-associated protein 2, Telomerase catalytic subunit, TP2) is encoded by the TERT (also known as CMM9, DKCA2, DKCB4, EST2, PFBMFT1, TCS1, TRT) gene (Gene ID 7015) in human. Telomerase reverse transcriptase (TERT) is the catalytic subunit of the telomerase responsible for adding TTAGGG repeats to the chromosome telomere ends. Cells with low or no telomerase expression lose telomere repeats during cell division, eventually resulting in cellular senescence. Most cancer cells, germ cells and embryonic stem cells express high levels of telomerase, thus contributing to pluripotency and immortality. In addition to its telomere maintenance function, telomerase also has a pro-survival role in cellular resistance against DNA damage and ensuing apoptosis. Most cancer cells are highly proliferative and express high levels of nuclear telomerase activity. Studies show that TERT shuttles from the nucleus into mitochondria upon oxidative stress induction in cancer cells. TERT mitochondrial localization helps prevent nuclear DNA damage by decreasing mitochondrial reactive oxygen species (ROS), accounting for high stress resistance especially among the cancer stem cells (CSCs) population. Following exposure to H2O2 or gamma-irradiation, cancer cells capable of excluding TERT from the nucleus display little or no DNA damage, while TERT nuclear retainment results in high DNA damage.

Specificity

Clone 7D5.2 targets a sequence toward the end of the C-terminal extension (CTE) region present in spliced isoforms 1 and 3, but not in isoforms 2 and 4 of human TERT reported by UniProt (O14746).

Immunogen

KLH-conjugated linear peptide corresponding to a sequence toward the end of the C-terminal extension (CTE) region of human TERT.

Application

Anti-TERT, clone 7D5.2 , Cat. No. MABE662, is a highly specific mouse monoclonal antibody that targets hTERT and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected TERT in human cerebral cortex, kidney, and tonsil tissue sections.

Western Blotting Analysis: 1 µg/mL from a representative lot detected TERT in 10 µg of Jurkat cell lysate.

Quality

Evaluated by Western Blotting in PC3 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected TERT in 10 µg of PC3 human prostate cancer cell lysate.

Target description

~140 kDa observed. 127.0/120.0 kDa (human isoform 1/3) calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: MABE14

Physical form

Format: Purified
Purified mouse IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service