Skip to Content
Merck
All Photos(1)

Key Documents

MABE369

Sigma-Aldrich

Anti-Suv4–20h2 Antibody, clone 6B9

clone 6B9, from rat

Synonym(s):

Histone-lysine N-methyltransferase SUV420H2, Lysine N-methyltransferase 5C, Suppressor of variegation 4-20 homolog 2, Su(var)4-20 homolog 2, Suv4-20h2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 423.00

CHF 423.00


Estimated to ship on06 May 2025



Select a Size

Change View
100 μG
CHF 423.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 423.00


Estimated to ship on06 May 2025


biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6B9, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Suppressor of variegation 4-20 homolog 2 (Suv4-20h2), also known as Lysine N-methyltransferase 5C (KMT5C), is a Histone-lysine N-methyltransferase that specifically trimethylates Lys-20 of histone H4. Suv4-20h2 mainly functions in pericentric heterochromatin regions, thereby playing a central role in the establishment of constitutive heterochromatin in these regions. In mammals, Histone H4 Lys-20 trimethylation (H4K20me3) is one of the most common chromatin modifications and it has been implicated in various biological processes, including heterochromatin formation, telomere function, imprinting, X inactivation, DNA damage repair and cancer progression.

Immunogen

GST-tagged recombinant protein corresponding to human Suv4-20h2.

Application

Detect the Suv4–20h2 protein using this Anti-Suv4–20h2, clone 6B9 validated for use in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blot in H9 human embryonic stem cell lysate.

Western Blot Analysis: 4.0 µg/mL of this antibody detected Suv4-20h2 in 10 µg of H9 human embryonic stem cell lysate.

Target description

~53 kDa observed. Uniprot describes 2 isoforms produced by alternative splicing at ~11 kDa and ~15 kDa

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
H9 human embryonic stem cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service