Skip to Content
Merck
All Photos(1)

Documents

MABE2028

Sigma-Aldrich

Anti-FAN1 Antibody, clone 1A11-2-A

Synonym(s):

EC:3.1.21, FANCD2/FANCI-associated nuclease 1, Fanconi-associated nuclease 1, Myotubularin-related protein 15, hFAN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A11-2-A, monoclonal

mol wt

calculated mol wt 114.23 kDa
observed mol wt ~115 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... FAN1(22909)

Specificity

Clone 1A11-2-A is a mouse monoclonal antibody that detects Fanconi-associated nuclease 1 (FAN1). It targets an epitope within the C-terminal region.

Immunogen

His-tagged recombinant fragment corresponding to 147 amino acids from the C-terminal region of human Fanconi-associated nuclease 1 (FAN1).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from wild-type HeLa cells.

Western Blotting Analysis: A 1:500 dilution of this antibody detected FAN1 in lysate from wild-type HeLa cells, but not in lysate form FAN1 knockout HeLa cells.

Tested Applications

Proximity Ligation Assay: A representative lot detected FAN1 in Proximity Ligation Assay (Porro, A., et al. (2021). Sci Adv. 7(31):eabf7906).

Western Blotting Analysis: A representative lot detected FAN1 in Western Blotting applications (Porro, A., et al. (2021). Sci Adv. 7(31):eabf7906).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Fanconi-associated nuclease 1 (UniProt: Q9Y2M0; also known as EC:3.1.21, FANCD2/FANCI-associated nuclease 1, hFAN1, Myotubularin-related protein 15) is encoded by the FAN1 (also known as KIAA1018, MTMR15) gene (Gene ID: 22909) in human. FAN1 is a nuclease that is required for the repair of DNA interstrand cross-links (ICL). It is recruited to the sites of DNA damage by monoubiquitinated FANCD2, an effector of ATR signaling. It acts as a 5′-3′ exonuclease that anchors at a cut end of DNA and cleaves DNA successively at every third nucleotide, allowing to excise an ICL from one strand through flanking incisions. It is specifically involved in repair of ICL-induced DNA breaks and is required for efficient homologous recombination. However, it is not involved in DNA double-strand breaks resection. It is shown to hydrolytically remove 5′-nucleotides successively from the 3′-hydroxy termini of 3′-hydroxy-terminated oligonucleotides. FAN1 is reported to interact with MLH1 via two adjacent MLH1-binding sites, referred to as MLH1-interacting protein (MIP) box and MLH1-interacting motif (MIM). This interaction is regulated during the cell cycle and is inhibited in G2/M phase following CDK1/2-mediated phosphorylation of serine 126 within the MIP box of FAN1. Disruption of the FAN1-MLH1 interaction is reported to cause hypersensitivity to DNA cross-linking agents and defective repair of slipped-CAG/CTG repeats. Two isoforms of FAN1 have been described that are produced by alternative splicing. (Ref.: Porro, A., et al. (2021). , Sci. Adv. 7; eabf7906; Thongthip, S., et al. (2016). Genes & Dev. 2016. 30: 645-659; Wang, R., et al. (2014). Science. 346(6213); 1127-1130; Mackay, C., et al. (2010). Cell. 142(1); 65-76).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service