Skip to Content
Merck
All Photos(1)

Key Documents

MABE1904

Sigma-Aldrich

Anti-phospho-ACD (Ser255) Antibody, clone 34B10

Synonym(s):

Adrenocortical dysplasia protein homolog;TPP1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CHF 144.00
100 μG
CHF 364.00

CHF 144.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
25 μG
CHF 144.00
100 μG
CHF 364.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

CHF 144.00


Estimated to ship on30 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

34B10, monoclonal

mol wt

calculated mol wt 57.73 kDa
observed mol wt ~60 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer255)

Gene Information

human ... ACD(65057)

General description

Adrenocortical dysplasia protein homolog (UniProt: A0A590TQL1; also known as TPP1, ACD) is encoded by the ACD (also known as hCG_27140) gene (Gene ID: 65057) in human. ACD (TPP1) is one of the shelterin components that is involved in both telomere protection and telomere length regulation. Shelterin is known to associate with arrays of double-stranded TTAGGG repeats added by telomerase and protects chromosome ends. Without this protection, telomeres are no longer hidden from the DNA damage surveillance and chromosome ends are inappropriately processed by DNA repair pathways. ACD/TPP1 contains an oligosaccharide binding-fold (OB-fold) domain that provides an interface to recruit telomerase. Within its OB-fold domain is a cluster of amino acids known as the TEL patch (TPP1-Glu-Leu) that contains an interaction interface with telomerase. In the G2/M phase, it interacts with the cell cycle regulator kinase NEK6 that phosphorylates a highly conserved serine 255 that is located in the intervening region between the telomerase recruiting OB-fold and the POT1 (Protection Of Telomeres 1) recruiting domain. POT1 is reported to negatively regulate ACD/TPP1 phosphorylation. Phosphorylation of serine 255 is shown to be important in the interaction between ACD/TPP1 and telomerase. (Ref.: Henslee, G., et al. (2021). Cold Spring Harb. Mol. Case Stud. 7(1); a005454; Hirai, Y., et al. (2016). Genes Cells. 21(8); 874-889).

Specificity

Clone 34B10 is a mouse monoclonal antibody that detects ACD (TPP1) phosphorylated on serine 255.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids surrounding phosphoserine 255 from the internal region of human ACD/TPP1.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293T cell lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected ACD/TPP1 phosphorylated on serine 255 in lysates from HEK293T cells simultaneously transfected with HA-NEK6 and FLAG-TPP1 plasmids, but not in lysates from mock transfected cells (HEK293T/pcDNA3) or cells transfected only with FLAG-TPP1.

Tested Applications

Western Blotting Analysis: A representative lot detected ACD in Western Blotting applications (Hirai, Y., et al. (2016). Genes Cells. 21(8):874-89).

Immunocytochemistry Analysis: A representative lot detected ACD in Immunocytochemistry applications (Hirai, Y., et al. (2016). Genes Cells. 21(8):874-89).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-phospho-ACD (Ser255), clone 34B10, Cat. No. MABE1904, is a mouse monoclonal antibody that detects ACD (TPP1) phosphorylated on serine 255 and is tested for use in Immunocytochemistry and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service