Skip to Content
Merck
All Photos(1)

Key Documents

MABE1831

Sigma-Aldrich

Anti-CSB (ERCC6) Antibody, clone ICSB-3H8

clone 1CSB-3H8, from mouse

Synonym(s):

DNA excision repair protein ERCC-6, ATP-dependent helicase ERCC6, Cockayne syndrome protein CSB

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CHF 163.00
100 μG
CHF 457.00

CHF 163.00


Estimated to ship on24 May 2025



Select a Size

Change View
25 μG
CHF 163.00
100 μG
CHF 457.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 163.00


Estimated to ship on24 May 2025


biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

1CSB-3H8, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ChIP: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ERCC6(2074)

Related Categories

General description

DNA excision repair protein ERCC-6 (UniProt: Q03468; also known as ATP-dependent helicase ERCC6, Cockayne syndrome protein CSB) is encoded by the ERCC6 (also known as CSB) gene (Gene ID: 2074) in human. CSB is a homodimeric nuclear protein that belongs to the SwItch/ Sucrose nonfermentable family of DNA-dependent ATPases and contains seven characteristic helicase motifs. It is a part of a complex that contains RNA pol I, TFIIH, and XPG, all of which are active in transcription and/or DNA repair. Hence, it is an essential factor involved in transcription-coupled nucleotide excision repair, which allows RNA polymerase II-blocking lesions to be rapidly removed from the transcribed strand of active genes. Upon DNA-binding, it locally modifies DNA conformation by wrapping the DNA around itself, thereby modifying the interface between stalled RNA polymerase II and DNA. It is shown to be essential for transcription-coupled repair complex formation. It recruits the CSA complex (DCX(ERCC8) complex), nucleotide excision repair proteins and EP300 to the at sites of RNA polymerase II-blocking lesions. Mutations in CSB (ERCC6) gene are known to cause age related macular degeneration and Cockayne syndrome B that is characterized by delayed neural development and severe progressive neurologic degeneration and mental retardation.

Specificity

Clone 1CSB-3H8 specifically detects DNA excision repair protein ERCC-6 in human cells. It targets an epitope with in 22 amino acids from the C-terminal region.

Immunogen

Ovalbumin-conjugated linear Peptide corresponding to 22 amino acids from the C-terminal region of human DNA excision repair protein ERCC-6.

Application

Anti-CSB (ERCC6), clone ICSB-3H8, Cat. No. MABE1831, is a highly specific mouse monoclonal antibody that targets DNA excision repair protein ERCC-6 and has been tested for use in Chromatin Immunoprecipitation (ChIP), Flow Cytometry, Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected CSB (ERCC6) in Immunocytochemistry applications (Bradsher, J., et. al. (2002). Mol Cell. 10(4):819-29).

Immunoprecipitation Analysis: A representative lot immunoprecipitated CSB (ERCC6) in Immunoprecipitation applications (Bradsher, J., et. al. (2002). Mol Cell. 10(4):819-29).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected CSB (ERCC6) in Chromatin Immunoprecipitation applications (Kristensen, U., et. al. (2013). Proc Natl Acad Sci USA. 110(25):E2261-70; Velez-Cruz, R., et. al. (2012). Proc Natl Acad Sci USA. 110(3):E212-20).

Western Blotting Analysis: A representative lot detected CSB (ERCC6) in Western Blotting applications (Bradsher, J., et. al. (2002). Mol Cell. 10(4):819-29).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Flow Cytometry in A431 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected CSB (ERCC6) in one million A431 cells.

Target description

168.42 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service