Skip to Content
Merck
All Photos(2)

Key Documents

MABE1783

Sigma-Aldrich

Anti-PP2AC alpha/beta Antibody, clone 11H12

clone 11H12, from mouse

Synonym(s):

Serine/threonine-protein phosphatase 2A catalytic subunit alpha, Serine/threonine-protein phosphatase 2A catalytic subunit beta, PP2A-alpha, PP2A-beta

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CHF 163.00
100 μL
CHF 398.00

CHF 163.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
CHF 163.00
100 μL
CHF 398.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 163.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11H12, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), bovine (based on 100% sequence homology), rabbit (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Serine/threonine-protein phosphatase 2A catalytic subunit alpha/beta isoforms (UniProt: P67775/P62714; also known as EC 3.1.3.16, PP2A-alpha/PP2A-beta) is encoded by the PPP2CA/PPP2CB gene (Gene ID: 5515/5516) in human. Protein phosphatase 2A (PP2A) is a ubiquitously expressed enzyme that contributes 0.3 to 1% of the total cellular protein in the mammalian cell and regulates various functions by dephosphorylating many critical molecules, such as Akt, p53, c-Myc, CK2, S6 kinase, and beta-catenin. It is the major phosphatase for microtubule-associated proteins (MAPs). PP2A acts as a pivotal cellular switch controlling important events in cell, such as cell cycle, apoptosis, and autophagy It suppresses mitogenic signals and is suggested as a tumor suppressor. Aberrant expression and mutations in the PP2AA and PP2AB subunits are reported in various human malignancies. PP2A is made up of a functional heterotrimeric complex, which includes a structural subunit (PP2AA), a regulatory subunit (PP2AB), and a catalytic subunit (PP2AC). The regulatory subunit determines its substrate specificity. Over 20 different interchangeable regulatory subunits have been described that allows it to act on a large number of substrates. PP2AC has multiple metal ion binding sites and binds two manganese ions per subunit. Two isoforms of PP2AC (alpha and beta) have been described that display high degree of homology. PP2AC alpha also has two isoforms (PP2AC alpha 1 and alpha 2). The PP2AC alpha 2 isoform is shown to be catalytically inactive because it lacks amino acids 193-246.

Specificity

Clone 11H12 detects human Serine/threonine-protein phosphatase 2A catalytic subunit alpha/beta isoforms (PP2AC alpha and beta). It targets an epitope within 15 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Protein Phosphatase 2A catalytic subunit (PP2AC). The immungen sequence is conserved between alpha and beta isoforms of PP2AC.

Application

Anti-PP2AC alpha/beta, clone 11H12, Cat. No. MABE1783, is a highly specific mouse monoclonal antibody that targets Serine/threonine-protein phosphatase 2A catalytic subunit alpha and beta isoforms and has been tested for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PP2AC alpha/beta in NIH3T3 mouse fibroblasts ectopically expressing N-terminally tagged human PP2A catalytic subunits (Courtesy of Dr. Stefan Schuechner and Dr. Egon Ogris, Medical University of Vienna, Austria).

Quality

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected PP2AC alpha/beta in A549 cell lysate.

Target description

~ 35 kDa observed; 35.58 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sathish K R Padi et al.
Nature, 625(7993), 195-203 (2023-12-21)
Progression through the cell cycle is controlled by regulated and abrupt changes in phosphorylation1. Mitotic entry is initiated by increased phosphorylation of mitotic proteins, a process driven by kinases2, whereas mitotic exit is achieved by counteracting dephosphorylation, a process driven

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service