Skip to Content
Merck
All Photos(2)

Key Documents

MABE121

Sigma-Aldrich

Anti-SMARCA4 Antibody, clone 20C3.2

clone 20C3.2, from mouse

Synonym(s):

Transcription activator BRG1, ATP-dependent helicase SMARCA4, BRG1-associated factor 190A, BAF190A, Mitotic growth and transcription activator, Protein BRG-1, Protein brahma homolog 1, SNF2-beta, SWI/SNF-related matrix-associated actin-dependent regulato

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 463.00

CHF 463.00


Estimated to ship on25 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1148

Request a Bulk Order

Select a Size

Change View
100 μG
CHF 463.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 463.00


Estimated to ship on25 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1148

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

20C3.2, monoclonal

species reactivity

rat, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SMARCA4(6597)

General description

Also known as Protein BRG1 and SNF2-beta, among other aliases, SMARCA4 a transcriptional activator belonging to the SNF2/RAD54 helicase family, and acts as a component of the SWI/SNF ATP-dependent chromatin remodeling complexes family. BRG1 contains 4 domains of interest; a bromo domain, a helicase ATP-binding domain, a helicase C-terminal domain, and a HAS domain. It is a component of several different complexes with distinctive roles such as the CREST-BRG1 complex which regulates the activation of promoters in a calcium-dependent manner, releasing repressor complexes and recruiting activator complexes. It is also a part of the nBAF and npBAF complexes which are involved in the chromatin remodeling of mononucleosomes and nucleosomal arrays. As a component of the SWI/SNF complex, SMARCA4 has a role in tumor suppression and has been shown to regulate cell cycle and differentiation when present in neoplasma.

Immunogen

GST-tagged recombinant protein corresponding to the human SMARCA4.

Application

Anti-SMARCA4 Antibody, clone 20C3.2 is a Mouse Monoclonal Antibody for detection of SMARCA4 also known as Transcription activator BRG1, SNF2-beta & has been validated in WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Western Blot Analysis: 0.5 µg/mL of this antibody detected SMARCA4 in PC-12 cell lysate.

Immunocytochemistry Analysis: 1 µg/mL of this antibody detected SMARCA4 in HeLa and A431 cells.

Quality

Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 0.5 µg/mL of this antibody detected SMARCA4 in 10 µg of HeLa nuclear extract.

Target description

~184 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoqing Lv et al.
Nature cell biology, 26(9), 1585-1596 (2024-07-31)
Cyclic GMP-AMP synthase (cGAS), a cytosolic DNA sensor that initiates a STING-dependent innate immune response, binds tightly to chromatin, where its catalytic activity is inhibited; however, mechanisms underlying cGAS recruitment to chromatin and functions of chromatin-bound cGAS (ccGAS) remain unclear.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service