Skip to Content
Merck
All Photos(1)

Key Documents

MABE1115

Sigma-Aldrich

Anti-Pax1 Antibody, clone 5A2

from rat

Synonym(s):

Paired box protein Pax-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2a

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Pax1(18503)

General description

Paired box protein Pax-1 (UniProt: P09084) is encoded by the Pax1 (also known as Pax-1) gene (Gene ID: 18503) in murine species. Pax-1 is a transcriptional activator that plays a role in the formation of segmented structures of the embryo and has important role in the normal development of the vertebral column. Mutations in Pax1 gene are associated with vertebral malformations. Undulated homozygous mice exhibits vertebral malformations along the entire rostro-caudal axis. Pax genes are vital for the specification of progenitor cells and for maintenance of progenitor cell fate, including that of cancer cells. Pax1 gene is silenced by methylation in ovarian and cervical cancers and may serve as a tumor suppressor gene. The methylation levels of Pax1 gene is significantly higher in tumor tissues compared to non-tumor paracancerous tissues. Hence, methylation status of Pax1 may serve as useful biomarkers for the detection of cervical cancer and is also a good prognostic indicator for oral squamous cell carcinoma. (Ref.: Huang J et al (2017). Int. J. Environ. Res. Public Health. 14(2). pii: E216).

Specificity

Clone 5A2 detects Paired box protein Pax-1 in murine tissues.

Immunogen

GST-tagged full-length murine recombinant paired box protein Pax-1.

Application

Detect Paired box protein Pax-1 using this rat monoclonal Anti-Pax1, clone 5A2, Cat. No. MABE1115-25UG, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A representative lot detected Pax1 in Western Blotting applications (Feederle, R., et. al. (2016). Monoclon Antib Immunodiagn Immunother. [Epub ahead of print]).

Immunohistochemistry Analysis: A representative lot detected Pax1 in Immunohistochemistry applications (Feederle, R., et. al. (2016). Monoclon Antib Immunodiagn Immunother. [Epub ahead of print]; Kist, R., et. al. (2014). PLoS Genet. 10(10):e1004709).

Immunohistochemistry Analysis: A 1:200 dilution from a representative lot detected Pax1 in Embryonic mouse head (E14.5) (Courtesy of Dr. Heiko Peters, Ph.D., Institute of Genetic Medicine, Newcastle University, United Kingdom).

Western Blotting Analysis: A 1:10 dilution from a representative lot detected Pax1 in d11.5 embryonic vertebral column tissue lysates (Courtesy of Dr. Heiko Peters, Ph.D., Institute of Genetic Medicine, Newcastle University, United Kingdom).

Quality

Evaluated by Western Blotting in mouse esophagus tissue lysate.
Western Blotting Analysis: 1 µg/mL of this antibody detected Pax1 in 10 µg of m ouse esophagus tissue lysate.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leyao Shen et al.
eLife, 11 (2022-03-10)
Cellular differentiation is associated with the acquisition of a unique protein signature that is essential to attain the ultimate cellular function and activity of the differentiated cell. This is predicted to result in unique biosynthetic demands that arise during differentiation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service