Skip to Content
Merck
All Photos(1)

Documents

MABD407

Sigma-Aldrich

Anti-SIP1 Antibody, clone 6E5

clone 6E5, 1 mg/mL, from mouse

Synonym(s):

Smad-interacting protein 1, SMADIP1, Zinc finger homeobox protein 1b, Zinc finger E-box-binding homeobox 2, SIP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6E5, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZEB2(9839)

General description

Smad-interacting protein 1 (SIP1, SMADIP1), also known as Zinc finger E-box-binding homeobox 2 (ZEB2), and Zinc finger homeobox protein 1b (ZFHX1B, ZFX1B), is a transcriptional inhibitor that binds to DNA sequence 5′-CACCT-3′ in different promoters and interacts with activated SMADs. ZEB2 has been shown to repress the transcription of the E-cadherin gene and induce epithelial-mesenchymal transition, a genetic program controlling cell migration during embryonic development and wound healing in vitro. It is suggested that ZEB2 contributes to tumor progression since it’s been demonstrated to protect cells from DNA damage-induced apoptosis. Recent studies have shown that ZEB2 is often observed in the cytoplasm in numerous cancer tissues, implying that its localization may be regulated in normal and tumor tissues. Mutations in this gene are also associated with Mowat-Wilson syndrome (MWIS), also known as Hirschsprung disease-mental syndrome.

Immunogen

Epitope: N-terminus
His-tagged recombinant protein corresponding to the N-terminus of Human SIP1.

Application

Anti-SIP1 Antibody, clone 6E5 is an antibody against SIP1 for use in western blotting, ICC & IHC.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This antibody has been shown to work in Immunocytochemistry and Immunohistochemistry applications (Oztas et al., 2010).

Quality

Evaluated by Western Blot in a SK-Hep1 cell lysate. 2ug/ml (1:500 dilution) of this antibody detected SIP-1 in a SK-Hep1 cell lysate.

Target description

~220 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service