Skip to Content
Merck
All Photos(3)

Key Documents

MABD171

Sigma-Aldrich

Anti-BMR2 Antibody, clone 3F6

ascites fluid, clone 3F6, from mouse

Synonym(s):

Bone morphogenetic protein receptor type-2BMP type-2 receptor, BMPR-2, Bone morphogenetic protein receptor type II, BMP type II receptor, BMPR-II

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 473.00

CHF 473.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
CHF 473.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 473.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3F6, monoclonal

species reactivity

mouse, monkey, human, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BMPR2(659)

General description

BMPR2 a member of the bone morphogenetic protein (BMP) receptor family of transmembrane serine/threonine kinases. The ligands of this receptor are BMPs, which are members of the TGF-β superfamily. BMPs are involved in endochondral bone formation and embryogenesis. These proteins transduce their signals through the formation of heteromeric complexes of two different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. Mutations in this gene have been associated with primary pulmonary hypertension, both familial and fenfluramine-associated, and with pulmonary venoocclusive disease.

Immunogen

Purified recombinant fragment of human BMR2 expressed in E. Coli.

Application

Anti-BMR2 Antibody, clone 3F6 is a mouse monoclonal antibody, validated for use in western blotting, Immunofluorescence & IHC.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected BMPR2 in Eca109 cells.

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected BMPR2 in muscle tissue and kidney cancer tissue.

Optimal working dilutions must be determined by end user.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blot in HeLa, A431, NIH/3T3, Cos7 and PC-12 cell lysates.

Western Blot Analysis: A 1:500-2,000 dilution of this antibody detected BMPR2 in HeLa, A431, NIH/3T3, Cos7 and PC-12 cell lysates.

Target description

~115 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa, A431, NIH/3T3, Cos7 and PC-12 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crosstalk between TGF-?/Smad3 and BMP/BMPR2 signaling pathways via miR-17-92 cluster in carotid artery restenosis.
Luo, T; Cui, S; Bian, C; Yu, X
Molecular and Cellular Biochemistry null
Jia Liu et al.
Scientific reports, 6, 23670-23670 (2016-04-01)
Bone morphogenetic proteins (BMPs) regulate hard tissue formation, including bone and tooth. Growth differentiation factor 5 (GDF5), a known BMP, is expressed in cartilage and regulates chondrogenesis, and mutations have been shown to cause osteoarthritis. Notably, GDF5 is also expressed
Xiaowei Nie et al.
Biomolecules, 11(12) (2021-12-25)
Pulmonary hypertension (PH) is characterized by vascular remodeling caused by marked proliferation of pulmonary artery smooth muscle cells (PASMCs). Andrographolide (ANDRO) is a potent anti-inflammatory agent which possesses antioxidant, and has anticarcinogenic activity. The present study examined potential therapeutic effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service