Skip to Content
Merck
All Photos(1)

Key Documents

MABC708

Sigma-Aldrich

Anti-SND1, clone 2D7 Antibody

ascites fluid, clone 2D7, from mouse

Synonym(s):

Staphylococcal nuclease domain-containing protein 1, 100 kDa coactivator, EBNA2 coactivator p100, Tudor domain-containing protein 11, p100 co-activator, SND1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 318.00

CHF 318.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
CHF 318.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 318.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2D7, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SND1(27044)

General description

Staphylococcal nuclease domain-containing protein 1, also known as 100 kDa coactivator or EBNA2 coactivator p100 or Tudor domain-containing protein 11 or p100 co-activator, and encoded by the gene name SND1 or TDRD11, plays an important role in PIM1 regulation of MYB acitivity and acts as a transcriptional activatior of EBNA-2. SND1 also interacts with EAV, NSP1,GTF2E1 and GTF2E2, and forms a ternary complex with STAT6 and POLR2A. SND1 ic localized to the cytoplasm and in IL-4 stimulated cells SND1 co-localizes with STAT6 in the nucleus. SND1 plays an important role in the assembly of STAT6 transcriptome and stimulates IL-4-dependent transcription by mediating interaction between STAT6 and CBP.

Immunogen

Purified recombinant fragment of human SND1 expressed in E. Coli

Application

Anti-SND1, clone 2D7 Antibody detects level of SND1 & has been published & validated for use in SND1.
Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers

Quality

Evaluated by Western Blotting in HeLa, Jurkat, HepG2, and SMMC-7721 cell lysates.

Western Blotting Analysis: A 1:500-1:2,000 dilution of this antibody detected SND1 in HeLa, Jurkat, HepG2, and SMMC-7721 cell lysates.

Target description

~102 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service