Skip to Content
Merck
All Photos(1)

Documents

MABC196

Sigma-Aldrich

Anti-Chitinase-3-like protein 1 (YKL-40) Antibody, clone mAY

clone mAY, from mouse

Synonym(s):

Chitinase-3-like protein 1, 39 kDa synovial protein, Cartilage glycoprotein 39, CGP-39, GP-39, hCGP-39, YKL-40

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mAY, monoclonal

species reactivity

human

technique(s)

activity assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CHI3L1(1116)

General description

YKL-40 (Chitinase-3-like protein 1; human cartilage glycoprotein 39) is a secreted glycoprotein belonging to the glycosyl hydrolase 18 (GH18) family. It is found primarily in chondrocytes, activated macrophages, and hepatocytes. The precise biological function of YKL-40 is still unknown; however, recent studies suggest that YKL-40 may play a role in cell differentiation and inflammation. YKL-40 is elevated in the sera of patients with inflammatory diseases or cancer, and it is currently being investigated as a potential biomarker for these conditions.

Immunogen

Recombinant protein corresponding to human Chitinase-3-like protein 1 (YKL-40).

Application

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected Chitinase-3-like protein 1 (YKL-40) in U87 cells (This protein blocks tumor angiogenesis tube formation and progression (Faibish, M., et al. (2011). Mol Cancer Ther. 10(5):742-751.)).

Activity Assay Analysis: A representative lot from an independent laboratory neutralized Chitinase-3-like protein 1 in a neutralization assay (Faibish, M., et al. (2011). Mol Cancer Ther. 10(5):742-751.).
Research Category
Cell Structure
Research Sub Category
Developmental Signaling
This Anti-Chitinase-3-like protein 1 (YKL-40) Antibody, clone mAY is validated for use in Western Blotting, IHC, Activity Assay for the detection of Chitinase-3-like protein 1 (YKL-40).

Quality

Evaluated by Western Blot in MG-63 conditioned media.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Chitinase-3-like protein 1 (YKL-40) in 10 µg of MG-63 conditioned media (This protein blocks tumor angiogenesis tube formation and progression (Faibish, M., et al. (2011). Mol Cancer Ther. 10(5):742-751.)).

Target description

~42 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
MG-63 conditioned media

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaori Watanabe et al.
International journal of oncology, 60(1) (2021-12-17)
The cancer‑stromal interaction has been demonstrated to promote tumor progression, and cancer-associated fibroblasts (CAFs), which are the main components of stromal cells, have attracted attention as novel treatment targets. Chitinase 3-like 1 (CHI3L1) is a chitinase-like protein, which affects cell proliferation
Dominic Schmid et al.
Journal of neurochemistry, 158(2), 522-538 (2021-03-19)
Recent technological advances in molecular diagnostics through liquid biopsies hold the promise to repetitively monitor tumor evolution and treatment response of brain malignancies without the need of invasive surgical tissue accrual. Here, we implemented a mass spectrometry-based protein analysis pipeline
Erika Morera et al.
In vitro cellular & developmental biology. Animal, 55(10), 838-853 (2019-09-05)
Epithelial to mesenchymal transition (EMT) is a developmental event that is hijacked in some diseases such as fibrosis and cancer. In cancer, EMT has been linked to increased invasion and metastasis and is generally associated with a poor prognosis. In
Jon P Joelsson et al.
ALTEX, 37(4), 545-560 (2020-05-26)
Azithromycin (AZM) is a broad-spectrum antibiotic widely used to treat infections. AZM also has been shown to have anti-inflammatory and immunomodulatory functions unrelated to its antibacterial activity that contribute to the effectiveness of this drug in chronic respiratory diseases. The
Jon P Joelsson et al.
ALTEX, 36(4), 634-642 (2019-06-19)
Mechanical ventilation (MV) is a life-saving therapy for critically ill patients, alleviating the work of breathing and supporting adequate gas exchange. However, MV can cause ventilator induced lung injury (VILI) by baro/volu- and atelectrauma, even lead to acute respiratory distress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service