Skip to Content
Merck
All Photos(2)

Documents

MABC1177

Sigma-Aldrich

Anti-Stat3b Antibody, clone 516 G10H9

clone 516 G10H9, from mouse

Synonym(s):

Signal transducer and activator of transcription 3b, Stat3b, Acute-phase response factor, spliced isoform 3, DNA-binding protein APRF, spliced isoform 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

516 G10H9, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... STAT3(6774)

General description

Signal transducer and activator of transcription 3 (UniProt P40763; also known as Acute-phase response factor, DNA-binding protein APRF) is encoded by the STAT3 (also known as ADMIO, APRF, HIES) gene (Gene ID 6774) in human. STAT proteins (1, 2, 3, 4, 5A, 5B, 6) constitute a family of proteins that mediate cytokine and growth factor receptors-initiated signaling events. Stat3 is activated upon phosphorylation on Y705 by Jak or receptor tyrosine kinases, leading to Stat3 homodimerization, nuclear translocation, and target genes transcription. Constitutively tyrosine-phosphorylated Stat3 is implicated in the initiation and progression of many types of cancers, while Stat3 gene knockout in mice is lethal at the early embryonic stage. Two major alternatively spliced Stat3 isoforms exist, the C-terminal end 55-amino acid sequence (a.a. 716-770) in the canonical alpha isoform (UniProt P40763-1) is truncated and replaced with a 7-amino acid sequence in the 722-amino acid beta isoform (UniProt P40763-3). Tyrosine-phosphorylated Stat3β homodimers are more stable and less susceptible to dephosphorylation, bind DNA more avidly and exhibit markedly prolonged nuclear retention than pStat3α homodimers.

Specificity

Clone 516 G10H9 detects the C-terminal end 7-a.a. (CT7) sequence present in human/mouse spliced isoform Stat3B (UniProt P40763-3/P42227-2), but not in human/mouse spliced isoform Stat3A (UniProt P40763-1/P42227-1) or human/mouse isoform Del-701 (UniProt P40763-2/P42227-3).

Immunogen

Epitope: Within Stat3beta C-terminal end 7 a.a. (CT7).
Linear peptide containing human Stat3b C-terminal end 7 a.a. (CT7).

Application

Anti-Stat3b Antibody, clone 516 G10H9 is an antibody against Stat3b for use in Western Blotting, Immunoprecipitation.
Research Category
Apoptosis & Cancer
Research Sub Category
Transcription Factors
Western Blotting Analysis: 1 µg/mL from a representative lot detected 0.3-100 ng of human Stat3β (a.a. 127–722), but not Stat3α (a.a. 127–770), GFP fusion protein (Courtesy of Dr. David J. Tweardy, Baylor College of Medicine, Houston, TX).
Western Blotting Analysis: A representative lot specifically detected Stat3β (a.a. 127–722) GFP fusion protein, but not Stat3α (a.a. 127–770) or Stat3βΔCT7 (a.a. 127–717) GFP fusion that lacks Stat3β C-terminal end 7-a.a. (CT7) sequence (Bharadwaj, U., et al. (2014). Cancers (Basel). 6(4):2012-2034).
Immunoprecipitation Analysis: A representative lot specifically immunoprecipitated human Stat3β (a.a. 127–722) GFP fusion protein, but not Stat3α (a.a. 127–770) GFP fusion protein, exogenously expressed in MEFs lacking endogenous murine Stat3 (Bharadwaj, U., et al. (2014). Cancers (Basel). 6(4):2012-2034).

Quality

Evalusated by Western Blotting of recombinant human Stat3β GFP fusion protein.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected 0.5 µg of human Stat3β (a.a. 127–722) GFP fusion protein.

Target description

83.13 kDa calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service