Skip to Content
Merck
All Photos(1)

Key Documents

MAB8582-M

Sigma-Aldrich

Mouse Anti-RSV Fusion Protein, clone 102-10B (KC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

antibody form

purified antibody

Quality Level

clone

102-10B
monoclonal

species reactivity

human, mouse

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG1κ

General description

A2

Specificity

Reacts with RSV fusion protein 1c.

Application

Epitope: all type B strains

Physical form

Liquid in 0.02M PB, 0.25M NaCl, 0.1% sodium azide pH 7.6.

Storage and Stability

Maintain at 2-8°C.

Disclaimer

This product is intended for use by IVD manufacturers.
Contact customer service for assistance: [email protected]
Minimum order quantity required to qualify. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service