Skip to Content
Merck
All Photos(1)

Key Documents

MAB4709

Sigma-Aldrich

Anti-Caspase 9 Antibody, clone 2-23

clone 2-23, Chemicon®, from mouse

Synonym(s):

ICE-LAP6, Mch-6, Apaf-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 414.00

CHF 414.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 414.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 414.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2-23, monoclonal

species reactivity

human, mouse

should not react with

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP9(842)

Specificity

Recognizes 46-48 kDa full-length procaspase-9, human recombinant caspase-9 and procaspase-9 from human cell lysates. Highly specific to caspase-9 and shows no cross-reaction with other caspase family members. Activation of procaspase-9 by Apaf-1 in the cytochrome c pathway requires proteolytic cleavage to generate active caspase-9. Once activated, capsase-9 initiates a caspase cascade involving the downstream executioner caspases 3, 6 and 7.

Immunogen

Recombinant human caspase-9 prodomain

Application

Anti-Caspase 9 Antibody, clone 2-23 is an antibody against Caspase 9 for use in IP, WB, IH(P).
Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Caspases

Enzymes & Biochemistry
Western blot: 1:1000 Immunohistochemistry (frozen and paraffin): 1:500 - 1:1000

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-443

Physical form

Format: Purified
Purified immunoglobulin by protein A. Liquid in PBS, pH 7.6, with 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uncoupling oxidative/energy metabolism with low sub chronic doses of 3-nitropropionic acid or iodoacetate in vivo produces striatal cell damage.
Rodriguez, E; Rivera, I; Astorga, S; Mendoza, E; Garcia, F; Hernandez-Echeagaray, E
International Journal of Biological Sciences null
Wanyuan Chen et al.
Oncology reports, 41(1), 67-76 (2018-10-27)
Pancreatic cancer (PC) is a lethal solid malignancy with resistance to traditional chemotherapy. Recently, considerable studies have demonstrated the ubiquitous antitumor properties of gene therapy mediated by the oncolytic vaccinia virus. The second mitochondrial‑derived activator of caspase (Smac) has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service